Assay Details
Gene Symbol: OR8K3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005202.1 | NP_001005202.1 | 1 | 836 | 110 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218304 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218348 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218305 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218326 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218286 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218354 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218311 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218329 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07808839 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218347 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07808850 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218339 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07808819 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218333 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07808827 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07808797 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07808837 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07808793 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218323 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218288 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218285 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218338 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218280 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07808842 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07808806 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07808830 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218307 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218281 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218336 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218316 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218344 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218318 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218345 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218335 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218337 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218289 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218334 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07808821 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07808861 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07808833 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07808858 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218346 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07808800 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07808853 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218295 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218290 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07808828 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218340 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218312 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218351 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218331 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218302 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218321 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218306 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218315 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218301 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218352 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218303 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218317 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218325 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218299 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218308 | - | - |
Agilent SurePrint G3 Human CGH Microarray 4x180K | 166243 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P36050 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_23_P36050 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 9300 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 33902 | - | - |
Agilent Human Genome CGH + SNP Microarray | A_16_P02447609 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P36050 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_23_P36050 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218338 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07808793 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218326 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218333 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07808800 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218331 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218295 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218316 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07808806 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218317 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218303 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218336 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218335 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218325 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218299 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218330 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218304 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218301 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07808828 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07808791 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218308 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218311 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218315 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218312 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218306 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07808833 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07808804 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218292 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07808830 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218293 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07808827 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07808797 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07808792 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07808819 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07808821 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07808830 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218325 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07808791 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218292 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07808792 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218304 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07808806 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07808821 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218326 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218293 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218303 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07808800 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218336 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218330 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218312 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07808828 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218306 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07808827 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07808797 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07808819 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218301 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218335 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218316 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07808804 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218295 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218338 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218333 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07808833 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218317 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218331 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07808793 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218311 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218315 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218308 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218299 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_23_P36050 | - | - |
Agilent Human Genome CGH Microarray 105A (G4412A) | A_16_P02447609 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P07808802 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P07808835 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P07808800 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P07808832 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P07808828 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P07808806 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P07808795 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P07808814 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P07808841 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P07808821 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P07808811 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P07808844 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P07808824 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P07808827 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P07808809 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P07808823 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P07808840 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P07808848 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P07808804 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P07808819 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P07808818 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P07808798 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P07808820 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P07808797 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P02447609 | - | - |
Agilent Whole Human Genome Microarray 4x44K G4112F | A_23_P36050 | - | - |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 194 | - | - |
Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 34851 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P02447609 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07808811 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07808839 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07808828 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07808857 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07808867 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07808820 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07808835 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07808821 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07808832 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07808793 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07808851 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07808841 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07808815 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07808844 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07808809 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07808806 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07808798 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07808849 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07808797 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07808824 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07808800 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07808795 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07808802 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16453279 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07808860 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07808853 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07808848 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07808840 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07808790 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16453276 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07808804 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07808863 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07808855 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07808818 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07808814 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07808865 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07808827 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P02447609 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | NM_001005202_at | - | - |
Affymetrix Human Gene 1.0 ST Array | 7939978 | - | - |
Affymetrix Human Gene 1.1 ST Array | 7939977 | - | - |
Affymetrix Human Genome U219 Array | 11742594_PM_at | - | - |
Affymetrix Human Genome U219 Array | 11742594_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | TC1100007632.hg.1 | ENST00000312711.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007632.hg.1 | ENST00000623750.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007632.hg.1 | ENST00000623845.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11005534.hg.1 | ENST00000312711.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11005534.hg.1 | NM_001005202.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11005534.hg.1 | TC11000450.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | TC1100007632.hg.1 | ENST00000312711.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007632.hg.1 | ENST00000623750.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007632.hg.1 | ENST00000623845.1 | Ensembl |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A046400 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A046400 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_1808262 | NM_001005202.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1808262 | NM_001005202.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1808262 | NM_001005202.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1808262 | NM_001005202.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1808262 | NM_001005202.1 | human_transcriptome |
Illumina HumanRef-8 v2 Expression BeadChip | GI_6933-S | - | - |
Illumina human-6 v2.0 expression beadchip | ILMN_1808262 | NM_001005202.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1808262 | NM_001005202.1 | human_transcriptome |
Illumina Sentrix Human-6 v2 Expression BeadChip | ILMN_6933 | - | - |
Illumina HumanRef-6 v2.0 expression beadchip | ILMN_6933 | - | - |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1808262 | NM_001005202.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1808262 | NM_001005202.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1808262 | NM_001005202.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1808262 | NM_001005202.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1808262 | NM_001005202.1 | human_transcriptome |
