Assay Details
| Gene Symbol: OR2L8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001963.1 | NP_001001963.1 | 1 | 343 | 103 | |
| GenBank mRNA | BC140751.1 | - | 1 | 424 | 9021619 | 103 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01002020.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00931011 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21005113 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00931009 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21005133 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00931001 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15204975 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21005126 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00931008 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21005139 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21005120 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21005140 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21005121 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21005110 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21005123 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21005138 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21005107 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00930989 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00931004 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21005136 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00930994 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21005124 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21005115 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00931000 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21005116 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21005108 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00931015 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00930991 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00931005 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21005117 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21005112 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00931002 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3370001 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_0215 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_0214 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 27798 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3369975 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3370001 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3370001 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21005139 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21005107 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21005136 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21005108 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15204975 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21005133 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21005121 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15204980 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15204977 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21005115 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00931000 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21005140 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00931008 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00930994 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21005116 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21005124 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00931001 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21005126 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00931005 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00931009 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21005123 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00931015 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00931004 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00931011 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00931004 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21005115 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21005123 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00931008 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00931005 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00930994 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21005136 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15204977 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21005140 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00931009 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00931015 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21005124 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15204980 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21005107 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21005121 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15204975 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21005116 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21005139 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21005108 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00931011 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21005133 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00931000 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00931001 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21005126 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P00931015 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P00931006 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P00930986 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P00931001 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P00931009 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P00931004 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P00931013 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P00931008 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P00931014 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P00931012 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P00931010 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P00931005 | - | - |
| Agilent Human 1B Microarray G4111A | A_32_P309422 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00931008 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00930993 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00931004 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15204975 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00930990 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00930988 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00931015 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00931006 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00931013 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00930992 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00930999 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00931005 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00930997 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00930995 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00930986 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00931001 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00931010 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | NM_001001963_at | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7911242 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 7911241 | - | - |
| Affymetrix Human Transcriptome Array 2.0 | PSR01031101.hg.1 | TC01002020.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01031101.hg.1 | ENST00000357191.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01031101.hg.1 | NM_001001963.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100190587.hg.1 | ENST00000357191.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100190587.hg.1 | ENST00000623922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100190587.hg.1 | NM_001001963.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100190587.hg.1 | ENST00000357191.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100190587.hg.1 | ENST00000623922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100190587.hg.1 | NM_001001963.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100190587.hg.1 | TC0100012334.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100190586.hg.1 | TC0100012334.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100190588.hg.1 | TC0100012334.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A042316 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A042312 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A042316 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A042312 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 611817 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_2129658 | BC140751.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_2129658 | NM_001004687.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_2129658 | BC137039.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_2129658 | BC137040.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_2129658 | NM_001001963.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1790301 | - | - |
| HumanHT-12 V4.0 expression beadchip | ILMN_2129658 | BC140751.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_2129658 | NM_001004687.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_2129658 | BC137039.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_2129658 | BC137040.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_2129658 | NM_001001963.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2129658 | BC140751.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2129658 | NM_001004687.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2129658 | BC137039.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2129658 | BC137040.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2129658 | NM_001001963.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2129658 | BC140751.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2129658 | NM_001004687.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2129658 | BC137039.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2129658 | BC137040.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2129658 | NM_001001963.1 | human_transcriptome |
| Illumina HumanRef-8 v2 Expression BeadChip | GI_12197-S | - | - |
| Illumina Sentrix Human-6 v2 Expression BeadChip | 1570253 | - | - |
| Illumina HumanRef-6 v2.0 expression beadchip | ILMN_12197 | - | - |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2129658 | BC140751.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2129658 | NM_001004687.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2129658 | BC137039.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2129658 | BC137040.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2129658 | NM_001001963.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2129658 | BC140751.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2129658 | NM_001004687.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2129658 | BC137039.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2129658 | BC137040.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2129658 | NM_001001963.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2129658 | BC140751.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2129658 | NM_001004687.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2129658 | BC137039.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2129658 | BC137040.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2129658 | NM_001001963.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1790301 | - | - |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2129658 | BC140751.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2129658 | NM_001004687.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2129658 | BC137039.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2129658 | BC137040.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2129658 | NM_001001963.1 | human_transcriptome |

OR2L8
Genomic Map