Assay Details
Gene Symbol: FABP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001444.2 | NP_001435.1 | 1 - 2 | 187 | 149 | |
GenBank mRNA | AK226036.1 | - | 1 - 2 | 136 | 149 | |
AK311856.1 | - | 1 - 2 | 134 | 149 | ||
BC019385.2 | - | 1 - 2 | 120 | 3862100 | 149 | |
BC070303.1 | - | 1 | 134 | 6496304 | 149 | |
BT007449.1 | - | 1 | 74 | 149 | ||
CN278427.1 | - | 1 - 2 | 307 | 149 | ||
M94856.1 | - | 1 - 2 | 122 | 149 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC08000518.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1100010987.hg.1 | ENST00000524732.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008080.hg.1 | ENST00000297258.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008080.hg.1 | ENST00000297258.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008080.hg.1 | ENST00000297258.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008080.hg.1 | ENST00000297258.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008080.hg.1 | ENST00000297258.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008080.hg.1 | ENST00000297258.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008080.hg.1 | ENST00000297258.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008080.hg.1 | ENST00000297258.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008080.hg.1 | ENST00000297258.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008080.hg.1 | ENST00000396359.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008080.hg.1 | ENST00000396359.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008080.hg.1 | ENST00000396359.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008080.hg.1 | ENST00000396359.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008080.hg.1 | ENST00000396359.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008080.hg.1 | ENST00000396359.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008080.hg.1 | ENST00000481695.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008080.hg.1 | ENST00000481695.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008080.hg.1 | ENST00000481695.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008080.hg.1 | ENST00000481695.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008080.hg.1 | ENST00000481695.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008080.hg.1 | ENST00000486269.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008080.hg.1 | ENST00000486269.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008080.hg.1 | ENST00000486269.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008080.hg.1 | ENST00000486269.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008080.hg.1 | NM_001444.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008080.hg.1 | NM_001444.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008080.hg.1 | NM_001444.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008080.hg.1 | NM_001444.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008080.hg.1 | NM_001444.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008080.hg.1 | NM_001444.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008080.hg.1 | NM_001444.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008080.hg.1 | NM_001444.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009141.hg.1 | ENST00000515543.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011505.hg.1 | ENST00000515537.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009659.hg.1 | ENST00000435729.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08007008.hg.1 | TC08000518.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR08007008.hg.1 | ENST00000297258.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08007008.hg.1 | NM_001444.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500008110.hg.1 | ENST00000559181.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011828.hg.1 | ENST00000510588.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08007012.hg.1 | TC08000518.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR08007012.hg.1 | ENST00000297258.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08007012.hg.1 | ENST00000396359.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08007012.hg.1 | NM_001444.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007872.hg.1 | ENST00000430910.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009737.hg.1 | ENST00000437960.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009737.hg.1 | ENST00000477993.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009737.hg.1 | ENST00000477993.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009737.hg.1 | ENST00000477993.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009737.hg.1 | ENST00000477993.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009737.hg.1 | ENST00000477993.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009737.hg.1 | NR_002935.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009737.hg.1 | NR_002935.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009737.hg.1 | NR_002935.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009737.hg.1 | NR_002935.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR08007006.hg.1 | TC08000518.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR08007006.hg.1 | ENST00000297258.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08007006.hg.1 | NM_001444.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300009240.hg.1 | ENST00000423401.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009240.hg.1 | ENST00000423401.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009240.hg.1 | ENST00000423401.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009240.hg.1 | ENST00000423401.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009240.hg.1 | ENST00000423401.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007236.hg.1 | ENST00000439042.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500008188.hg.1 | ENST00000559487.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010933.hg.1 | ENST00000517670.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010933.hg.1 | ENST00000517670.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010933.hg.1 | ENST00000518880.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010933.hg.1 | ENST00000518880.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010987.hg.1 | ENST00000524732.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009659.hg.1 | ENST00000435729.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009141.hg.1 | ENST00000515543.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010933.hg.1 | ENST00000517670.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010933.hg.1 | ENST00000517670.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010933.hg.1 | ENST00000518880.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010933.hg.1 | ENST00000518880.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500008188.hg.1 | ENST00000559487.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007872.hg.1 | ENST00000430910.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100182838.hg.1 | ENST00000524732.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100182838.hg.1 | ENST00000524732.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100182838.hg.1 | TC1100010987.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0500011828.hg.1 | ENST00000510588.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007236.hg.1 | ENST00000439042.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009240.hg.1 | ENST00000423401.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009240.hg.1 | ENST00000423401.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009240.hg.1 | ENST00000423401.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009240.hg.1 | ENST00000423401.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009240.hg.1 | ENST00000423401.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800155498.hg.1 | ENST00000297258.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800155498.hg.1 | NM_001444.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800155498.hg.1 | ENST00000297258.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800155498.hg.1 | NM_001444.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800155498.hg.1 | TC0800008080.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0800155501.hg.1 | ENST00000297258.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800155501.hg.1 | NM_001444.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800155501.hg.1 | ENST00000297258.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800155501.hg.1 | NM_001444.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800155501.hg.1 | TC0800008080.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1100182839.hg.1 | TC1100010987.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0700009737.hg.1 | ENST00000437960.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009737.hg.1 | ENST00000477993.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009737.hg.1 | ENST00000477993.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009737.hg.1 | ENST00000477993.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009737.hg.1 | ENST00000477993.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009737.hg.1 | ENST00000477993.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009737.hg.1 | NR_002935.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009737.hg.1 | NR_002935.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009737.hg.1 | NR_002935.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009737.hg.1 | NR_002935.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500008110.hg.1 | ENST00000559181.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800155500.hg.1 | ENST00000297258.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800155500.hg.1 | NM_001444.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800155500.hg.1 | ENST00000297258.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800155500.hg.1 | NM_001444.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800155500.hg.1 | TC0800008080.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0800155505.hg.1 | ENST00000297258.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800155505.hg.1 | ENST00000396359.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800155505.hg.1 | ENST00000481695.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800155505.hg.1 | ENST00000486269.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800155505.hg.1 | NM_001444.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800155505.hg.1 | ENST00000297258.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800155505.hg.1 | ENST00000396359.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800155505.hg.1 | ENST00000481695.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800155505.hg.1 | ENST00000486269.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800155505.hg.1 | NM_001444.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800155505.hg.1 | TC0800008080.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0500011505.hg.1 | ENST00000515537.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008080.hg.1 | ENST00000297258.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008080.hg.1 | ENST00000297258.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008080.hg.1 | ENST00000297258.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008080.hg.1 | ENST00000297258.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008080.hg.1 | ENST00000297258.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008080.hg.1 | ENST00000297258.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008080.hg.1 | ENST00000297258.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008080.hg.1 | ENST00000297258.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008080.hg.1 | ENST00000297258.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008080.hg.1 | ENST00000396359.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008080.hg.1 | ENST00000396359.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008080.hg.1 | ENST00000396359.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008080.hg.1 | ENST00000396359.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008080.hg.1 | ENST00000396359.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008080.hg.1 | ENST00000396359.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008080.hg.1 | ENST00000481695.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008080.hg.1 | ENST00000481695.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008080.hg.1 | ENST00000481695.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008080.hg.1 | ENST00000481695.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008080.hg.1 | ENST00000481695.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008080.hg.1 | ENST00000486269.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008080.hg.1 | ENST00000486269.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008080.hg.1 | ENST00000486269.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008080.hg.1 | ENST00000486269.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008080.hg.1 | NM_001444.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008080.hg.1 | NM_001444.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008080.hg.1 | NM_001444.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008080.hg.1 | NM_001444.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008080.hg.1 | NM_001444.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008080.hg.1 | NM_001444.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008080.hg.1 | NM_001444.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008080.hg.1 | NM_001444.2 | Refseq |
