Assay Details
| Gene Symbol: EIF2S2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001316363.1 | NP_001303292.1 | 6 - 7 | 924 | 151 | |
| NM_003908.4 | NP_003899.2 | 6 - 7 | 933 | 151 | ||
| XM_017028118.1 | XP_016883607.1 | 6 - 7 | 1095 | 151 | ||
| GenBank mRNA | AB451237.1 | - | 5 - 6 | 687 | 151 | |
| AB451362.1 | - | 5 - 6 | 687 | 151 | ||
| AK313025.1 | - | 6 - 7 | 804 | 151 | ||
| BC000461.2 | - | 6 - 7 | 823 | 2822324 | 151 | |
| BC000934.2 | - | 6 - 7 | 798 | 3447202 | 151 | |
| BC007888.2 | - | 5 - 6 | 614 | 4120898 | 151 | |
| BT009795.1 | - | 5 - 6 | 687 | 151 | ||
| BX429051.2 | - | 6 - 7 | 770 | 151 | ||
| CR456734.1 | - | 5 - 6 | 687 | 151 | ||
| M29536.1 | - | 6 - 7 | 803 | 151 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

EIF2S2
Genomic Map