Assay Details
Gene Symbol: OR5B3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005469.1 | NP_001005469.1 | 1 | 248 | 136 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28226831 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28226849 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28226858 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28226863 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28226815 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07816962 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28226838 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28226867 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07816919 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07816936 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16454759 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07816948 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28226809 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28226835 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07816969 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07816951 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16454761 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28226812 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07816968 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07816957 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28226852 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07816956 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28226844 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28226834 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28226803 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07816950 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16454765 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28226832 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07816964 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28226817 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28226801 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28226833 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07816959 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28226814 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28226862 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28226808 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28226813 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07816960 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28226865 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28226854 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28226816 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07816945 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07816949 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28226840 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28226850 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07816916 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3352263 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_0654 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_0653 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 8602 | - | - |
Agilent Human Genome CGH + SNP Microarray | A_16_P02450259 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3352263 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_33_P3352263 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07816959 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28226835 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28226862 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28226850 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07816962 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07816948 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07816945 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28226858 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07816964 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07816960 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16454763 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07816949 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07816957 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28226833 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16454761 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16454765 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07816955 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07816968 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28226854 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07816969 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28226838 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07816971 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28226849 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16454759 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07816950 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28226865 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28226832 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28226867 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28226844 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07816955 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07816949 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28226844 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28226858 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28226833 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28226862 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28226867 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07816950 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16454765 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07816971 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07816964 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28226850 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28226849 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28226832 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16454759 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16454763 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28226838 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28226865 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07816969 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07816962 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07816957 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28226854 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07816945 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07816960 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07816948 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28226835 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16454761 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07816968 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07816959 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P07816963 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P07816941 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P07816953 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P07816946 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P07816971 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P07816966 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P07816956 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P07816970 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P07816947 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P07816958 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P07816967 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P07816938 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P07816949 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P07816943 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P07816961 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P07816951 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07816971 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07816956 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07816946 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07816942 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07816933 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07816967 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07816966 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07816951 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07816961 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16454768 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07816923 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16454765 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07816958 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07816926 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07816919 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07816920 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07816970 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07816949 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07816963 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07816940 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07816928 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07816937 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07816953 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07816914 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07816943 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07816917 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07816930 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07816935 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | NM_001005469_at | - | - |
Affymetrix Human Gene 1.0 ST Array | 7948325 | - | - |
Affymetrix Human Gene 1.1 ST Array | 7948324 | - | - |
Affymetrix Human Genome U219 Array | 11742575_at | - | - |
Affymetrix Human Genome U219 Array | 11742575_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A045209 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A045209 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_1693693 | NM_001005469.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1693693 | NM_001005469.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1693693 | NM_001005469.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1693693 | NM_001005469.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1693693 | NM_001005469.1 | human_transcriptome |
Illumina HumanRef-8 v2 Expression BeadChip | GI_2326-S | - | - |
Illumina human-6 v2.0 expression beadchip | ILMN_1693693 | NM_001005469.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1693693 | NM_001005469.1 | human_transcriptome |
Illumina Sentrix Human-6 v2 Expression BeadChip | ILMN_2326 | - | - |
Illumina HumanRef-6 v2.0 expression beadchip | ILMN_2326 | - | - |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1693693 | NM_001005469.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1693693 | NM_001005469.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1693693 | NM_001005469.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1693693 | NM_001005469.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1693693 | NM_001005469.1 | human_transcriptome |
