Assay Details
Gene Symbol: NCR3LG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202439.2 | NP_001189368.1 | 4 - 5 | 1059 | 116 | |
XM_011520072.2 | XP_011518374.1 | 4 - 5 | 3013 | 116 | ||
XM_011520073.2 | XP_011518375.1 | 4 - 5 | 2759 | 116 | ||
XM_011520074.2 | XP_011518376.1 | 4 - 5 | 1376 | 116 | ||
XM_011520075.2 | XP_011518377.1 | 4 - 5 | 2665 | 116 | ||
GenBank mRNA | BC136797.1 | - | 4 - 5 | 922 | 116 | |
BC136800.1 | - | 4 - 5 | 922 | 116 | ||
BX537685.1 | - | 4 - 5 | 1051 | 116 | ||
CR749521.1 | - | 4 - 5 | 1059 | 116 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | nucleobindin 2 | NUCB2 |
HEL-S-109, NEFA |
-18.9kb | Sense | Chr. 11 - 17260340 - 17331881 on NCBI Build GRCh38 |
Downstream | ATP binding cassette subfamily C member 8 | ABCC8 |
ABC36, HHF1, HI, HRINS, MRP8, PHHI, SUR, SUR1, SUR1delta2, TNDM2 |
15.6kb | Anti-sense | Chr. 11 - 17392885 - 17476870 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC11000229.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1100006966.hg.1 | ENST00000338965.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006966.hg.1 | ENST00000338965.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006966.hg.1 | ENST00000338965.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006966.hg.1 | ENST00000338965.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006966.hg.1 | ENST00000338965.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006966.hg.1 | ENST00000338965.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006966.hg.1 | ENST00000338965.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006966.hg.1 | ENST00000338965.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006966.hg.1 | ENST00000338965.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006966.hg.1 | ENST00000338965.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006966.hg.1 | ENST00000338965.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006966.hg.1 | ENST00000338965.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006966.hg.1 | ENST00000530403.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006966.hg.1 | ENST00000530403.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006966.hg.1 | ENST00000530403.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006966.hg.1 | ENST00000530403.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006966.hg.1 | ENST00000530403.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006966.hg.1 | ENST00000530403.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006966.hg.1 | ENST00000530403.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006966.hg.1 | ENST00000530403.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006966.hg.1 | NM_001202439.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006966.hg.1 | NM_001202439.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006966.hg.1 | NM_001202439.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006966.hg.1 | NM_001202439.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006966.hg.1 | NM_001202439.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006966.hg.1 | NM_001202439.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006966.hg.1 | NM_001202439.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006966.hg.1 | NM_001202439.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006966.hg.1 | NM_001202439.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006966.hg.1 | NM_001202439.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006966.hg.1 | NM_001202439.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012769.hg.1 | ENST00000461341.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012769.hg.1 | ENST00000461341.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012769.hg.1 | ENST00000461341.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012769.hg.1 | ENST00000461341.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012769.hg.1 | ENST00000461341.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012769.hg.1 | ENST00000461341.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012769.hg.1 | ENST00000461341.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012769.hg.1 | ENST00000461341.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012769.hg.1 | ENST00000461341.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012769.hg.1 | ENST00000461341.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012769.hg.1 | ENST00000461341.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012769.hg.1 | ENST00000461341.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012769.hg.1 | ENST00000461341.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012769.hg.1 | ENST00000461341.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012769.hg.1 | ENST00000461341.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012769.hg.1 | ENST00000461341.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012769.hg.1 | ENST00000461341.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012769.hg.1 | ENST00000461341.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012769.hg.1 | ENST00000461341.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012769.hg.1 | ENST00000461341.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012769.hg.1 | ENST00000461341.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012769.hg.1 | ENST00000461341.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012769.hg.1 | ENST00000465363.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012769.hg.1 | ENST00000465363.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012769.hg.1 | ENST00000465363.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012769.hg.1 | ENST00000471823.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012769.hg.1 | ENST00000471823.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012769.hg.1 | ENST00000471823.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012769.hg.1 | ENST00000471823.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012769.hg.1 | ENST00000482639.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012769.hg.1 | ENST00000482639.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012769.hg.1 | ENST00000482639.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012769.hg.1 | ENST00000482639.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012769.hg.1 | ENST00000482639.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11003008.hg.1 | ENST00000338965.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11003008.hg.1 | NM_001202439.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11003008.hg.1 | TC11000229.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1100149034.hg.1 | ENST00000338965.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100149034.hg.1 | NM_001202439.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100149034.hg.1 | ENST00000338965.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100149034.hg.1 | NM_001202439.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100149034.hg.1 | TC1100006966.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1100149035.hg.1 | ENST00000338965.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100149035.hg.1 | ENST00000530403.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100149035.hg.1 | NM_001202439.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100149035.hg.1 | ENST00000338965.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100149035.hg.1 | ENST00000530403.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100149035.hg.1 | NM_001202439.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100149035.hg.1 | TC1100006966.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1100149036.hg.1 | ENST00000338965.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100149036.hg.1 | NM_001202439.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100149036.hg.1 | ENST00000338965.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100149036.hg.1 | NM_001202439.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100149036.hg.1 | TC1100006966.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1100006966.hg.1 | ENST00000338965.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006966.hg.1 | ENST00000338965.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006966.hg.1 | ENST00000338965.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006966.hg.1 | ENST00000338965.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006966.hg.1 | ENST00000338965.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006966.hg.1 | ENST00000338965.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006966.hg.1 | ENST00000338965.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006966.hg.1 | ENST00000338965.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006966.hg.1 | ENST00000338965.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006966.hg.1 | ENST00000338965.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006966.hg.1 | ENST00000338965.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006966.hg.1 | ENST00000338965.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006966.hg.1 | ENST00000530403.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006966.hg.1 | ENST00000530403.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006966.hg.1 | ENST00000530403.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006966.hg.1 | ENST00000530403.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006966.hg.1 | ENST00000530403.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006966.hg.1 | ENST00000530403.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006966.hg.1 | ENST00000530403.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006966.hg.1 | ENST00000530403.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006966.hg.1 | NM_001202439.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006966.hg.1 | NM_001202439.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006966.hg.1 | NM_001202439.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006966.hg.1 | NM_001202439.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006966.hg.1 | NM_001202439.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006966.hg.1 | NM_001202439.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006966.hg.1 | NM_001202439.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006966.hg.1 | NM_001202439.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006966.hg.1 | NM_001202439.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006966.hg.1 | NM_001202439.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006966.hg.1 | NM_001202439.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100149033.hg.1 | ENST00000338965.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100149033.hg.1 | ENST00000530403.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100149033.hg.1 | NM_001202439.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100149033.hg.1 | ENST00000338965.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100149033.hg.1 | ENST00000530403.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100149033.hg.1 | NM_001202439.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100149033.hg.1 | TC1100006966.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1100149032.hg.1 | ENST00000338965.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100149032.hg.1 | ENST00000530403.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100149032.hg.1 | NM_001202439.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100149032.hg.1 | ENST00000338965.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100149032.hg.1 | ENST00000530403.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100149032.hg.1 | NM_001202439.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100149032.hg.1 | TC1100006966.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0300012769.hg.1 | ENST00000461341.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012769.hg.1 | ENST00000461341.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012769.hg.1 | ENST00000461341.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012769.hg.1 | ENST00000461341.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012769.hg.1 | ENST00000461341.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012769.hg.1 | ENST00000461341.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012769.hg.1 | ENST00000461341.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012769.hg.1 | ENST00000461341.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012769.hg.1 | ENST00000461341.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012769.hg.1 | ENST00000461341.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012769.hg.1 | ENST00000461341.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012769.hg.1 | ENST00000461341.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012769.hg.1 | ENST00000461341.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012769.hg.1 | ENST00000461341.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012769.hg.1 | ENST00000461341.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012769.hg.1 | ENST00000461341.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012769.hg.1 | ENST00000461341.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012769.hg.1 | ENST00000461341.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012769.hg.1 | ENST00000461341.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012769.hg.1 | ENST00000461341.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012769.hg.1 | ENST00000461341.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012769.hg.1 | ENST00000461341.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012769.hg.1 | ENST00000465363.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012769.hg.1 | ENST00000465363.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012769.hg.1 | ENST00000465363.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012769.hg.1 | ENST00000471823.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012769.hg.1 | ENST00000471823.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012769.hg.1 | ENST00000471823.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012769.hg.1 | ENST00000471823.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012769.hg.1 | ENST00000482639.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012769.hg.1 | ENST00000482639.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012769.hg.1 | ENST00000482639.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012769.hg.1 | ENST00000482639.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012769.hg.1 | ENST00000482639.2 | Ensembl |
