Assay Details
| Gene Symbol: LOC285766 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_126020.1 | 6 - 7 | 1709 | 131 | ||
| GenBank mRNA | AK097546.1 | - | 6 - 7 | 1709 | 131 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | long intergenic non-protein coding RNA 266-3 | LINC00266-3 |
C6orf88, NCRNA00266-3, NCRNA00266C, dJ24O22.2 |
-34.2kb | Sense | Chr. 6 - 140264 - 148159 on NCBI Build GRCh38 |
| Downstream | dual specificity phosphatase 22 | DUSP22 |
JKAP, JSP-1, JSP1, LMW-DSP2, LMWDSP2, MKP-x, MKPX, VHX |
107.2kb | Anti-sense | Chr. 6 - 292057 - 351355 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC285766
Genomic Map