Assay Details
Gene Symbol: LOC285766 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_126020.1 | 7 - 8 | 1822 | 153 | ||
GenBank mRNA | AK097546.1 | - | 7 - 8 | 1822 | 153 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | long intergenic non-protein coding RNA 266-3 | LINC00266-3 |
C6orf88, NCRNA00266-3, NCRNA00266C, dJ24O22.2 |
-33.3kb | Sense | Chr. 6 - 140264 - 148159 on NCBI Build GRCh38 |
Downstream | dual specificity phosphatase 22 | DUSP22 |
JKAP, JSP-1, JSP1, LMW-DSP2, LMWDSP2, MKP-x, MKPX, VHX |
109.6kb | Anti-sense | Chr. 6 - 292057 - 351355 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24686599 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04500242 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24686550 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24686594 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24686583 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24686542 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24686614 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04500302 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24686676 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24686582 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04500287 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24686548 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04500276 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04500284 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04500277 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24686672 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04500283 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04500375 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04500376 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04500308 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04500274 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24686551 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15775993 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15776003 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15775999 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24686545 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24686549 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04500262 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04500290 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24686580 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04500265 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24686544 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04500313 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24686673 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04500285 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04500300 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04500261 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04500372 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04500294 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15776025 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04500377 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04500282 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04500289 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04500270 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04500281 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24686541 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15775992 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24686557 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24686553 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24686530 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04500296 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15775980 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15775974 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04500268 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04500260 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04500293 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04500309 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04500267 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04500306 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15776002 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24686601 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04500288 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04500286 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24686539 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24686547 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04500244 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24686682 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04500264 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04500273 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04500368 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04500246 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24686574 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04500240 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04500303 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04500311 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24686533 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04500369 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04500305 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24686563 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24686571 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24686543 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24686568 | - | - |
Agilent SurePrint G3 Human CGH Microarray 4x180K | 161317 | - | - |
Agilent SurePrint G3 Human CGH Microarray 4x180K | 27723 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 772106 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 754868 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 345354 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 316925 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 408022 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 381970 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 54312 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 371151 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 131142 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 417662 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 400731 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 265853 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 252371 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 141008 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 14055 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 386662 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 76813 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 377230 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 370507 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 186119 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 229382 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 380543 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 313585 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 70315 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 411355 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 140002 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 239059 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 205251 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 15534 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3773200 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 52028 | - | - |
Agilent Human Genome CGH + SNP Microarray | A_16_P01406770 | - | - |
Agilent Human Genome CGH + SNP Microarray | A_16_P37488478 | - | - |
Agilent Human Genome CGH + SNP Microarray | A_16_P01406770 | - | - |
Agilent Human Genome CGH + SNP Microarray | A_16_P37488518 | - | - |
Agilent Human Genome CGH + SNP Microarray | A_16_P01406746 | - | - |
Agilent Human Genome CGH + SNP Microarray | A_18_P24720843 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_33_P3773200 | - | - |
Agilent Human Genome CGH Microarray 105A (G4412A) | A_16_P37488478 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P37488478 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P01406770 | - | - |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 7334 | - | - |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 7420 | - | - |
Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 68142 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P01406770 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P37488524 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P24720843 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P15519057 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P17420757 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P01406773 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P15516111 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P01406740 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P17420728 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P15516752 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P01406746 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P17420704 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_18_P24720843 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P01406746 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P01406770 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P37488524 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_18_P15516221 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix HT HG-U133+ PM Array Plate | 1564253_PM_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | TC0600010359.hg.1 | ENST00000580119.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010359.hg.1 | ENST00000580119.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010359.hg.1 | ENST00000580119.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010359.hg.1 | ENST00000580119.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010359.hg.1 | ENST00000580119.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010359.hg.1 | ENST00000580119.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010359.hg.1 | ENST00000580119.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010359.hg.1 | ENST00000580119.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010359.hg.1 | NR_126020.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010359.hg.1 | NR_126020.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010359.hg.1 | NR_126020.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010359.hg.1 | NR_126020.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010359.hg.1 | NR_126020.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010359.hg.1 | NR_126020.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010359.hg.1 | NR_126020.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010359.hg.1 | NR_126020.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600170625.hg.1 | ENST00000580119.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600170625.hg.1 | NR_126020.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600170625.hg.1 | ENST00000580119.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600170625.hg.1 | NR_126020.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600170625.hg.1 | TC0600010359.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0600010359.hg.1 | ENST00000580119.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010359.hg.1 | ENST00000580119.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010359.hg.1 | ENST00000580119.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010359.hg.1 | ENST00000580119.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010359.hg.1 | ENST00000580119.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010359.hg.1 | ENST00000580119.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010359.hg.1 | ENST00000580119.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010359.hg.1 | ENST00000580119.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010359.hg.1 | NR_126020.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010359.hg.1 | NR_126020.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010359.hg.1 | NR_126020.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010359.hg.1 | NR_126020.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010359.hg.1 | NR_126020.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010359.hg.1 | NR_126020.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010359.hg.1 | NR_126020.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010359.hg.1 | NR_126020.1 | Refseq |
Human Genome U133 Plus 2.0 Array | 1564253_at | - | - |
Human Genome U133 Plus 2.0 Array | 1564253_PM_at | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1564253_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1564253_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1564253_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1564253_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1564253_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1564253_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1564253_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1564253_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1564253_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1564253_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1564253_at5 | - | - |
