Assay Details
Gene Symbol: TUBG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001070.4 | NP_001061.2 | 3 - 4 | 734 | 157 | |
GenBank mRNA | AK313339.1 | - | 3 - 4 | 403 | 157 | |
BC000619.2 | - | 3 - 4 | 398 | 3345973 | 157 | |
BT019931.1 | - | 3 - 4 | 336 | 157 | ||
CR407642.1 | - | 3 - 4 | 336 | 157 | ||
M61764.1 | - | 3 - 4 | 360 | 157 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC17000531.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1700007904.hg.1 | ENST00000251413.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007904.hg.1 | ENST00000251413.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007904.hg.1 | ENST00000251413.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007904.hg.1 | ENST00000251413.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007904.hg.1 | ENST00000251413.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007904.hg.1 | ENST00000251413.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007904.hg.1 | ENST00000251413.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007904.hg.1 | ENST00000251413.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007904.hg.1 | ENST00000251413.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007904.hg.1 | ENST00000251413.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007904.hg.1 | ENST00000251413.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007904.hg.1 | ENST00000251413.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007904.hg.1 | ENST00000251413.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007904.hg.1 | ENST00000251413.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007904.hg.1 | ENST00000588056.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007904.hg.1 | ENST00000588056.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007904.hg.1 | ENST00000588056.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007904.hg.1 | ENST00000588056.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007904.hg.1 | ENST00000588056.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007904.hg.1 | ENST00000588056.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007904.hg.1 | ENST00000589613.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007904.hg.1 | ENST00000589613.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007904.hg.1 | ENST00000589613.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007904.hg.1 | ENST00000589613.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007904.hg.1 | ENST00000589688.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007904.hg.1 | ENST00000589688.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007904.hg.1 | ENST00000589688.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007904.hg.1 | ENST00000589688.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007904.hg.1 | ENST00000589688.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007904.hg.1 | ENST00000589688.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007904.hg.1 | ENST00000589688.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007904.hg.1 | ENST00000589688.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007904.hg.1 | ENST00000589688.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007904.hg.1 | ENST00000589688.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007904.hg.1 | ENST00000589688.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007904.hg.1 | ENST00000589688.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007904.hg.1 | ENST00000589688.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007904.hg.1 | ENST00000591509.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007904.hg.1 | ENST00000591509.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007904.hg.1 | ENST00000591509.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007904.hg.1 | ENST00000591509.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007904.hg.1 | ENST00000591509.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007904.hg.1 | ENST00000591509.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007904.hg.1 | ENST00000591509.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007904.hg.1 | ENST00000591509.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007904.hg.1 | ENST00000591509.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007904.hg.1 | ENST00000591509.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007904.hg.1 | NM_001070.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007904.hg.1 | NM_001070.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007904.hg.1 | NM_001070.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007904.hg.1 | NM_001070.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007904.hg.1 | NM_001070.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007904.hg.1 | NM_001070.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007904.hg.1 | NM_001070.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007904.hg.1 | NM_001070.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007904.hg.1 | NM_001070.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007904.hg.1 | NM_001070.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007904.hg.1 | NM_001070.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007904.hg.1 | NM_001070.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007904.hg.1 | NM_001070.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007904.hg.1 | NM_001070.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007904.hg.1 | NM_001070.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17006437.hg.1 | ENST00000251413.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17006437.hg.1 | NM_001070.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17006437.hg.1 | TC17000531.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1700157589.hg.1 | ENST00000251413.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700157589.hg.1 | ENST00000588056.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700157589.hg.1 | ENST00000589688.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700157589.hg.1 | ENST00000591509.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700157589.hg.1 | NM_001070.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700157589.hg.1 | ENST00000251413.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700157589.hg.1 | ENST00000588056.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700157589.hg.1 | ENST00000589688.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700157589.hg.1 | ENST00000591509.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700157589.hg.1 | NM_001070.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700157589.hg.1 | TC1700007904.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1700007904.hg.1 | ENST00000251413.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007904.hg.1 | ENST00000251413.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007904.hg.1 | ENST00000251413.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007904.hg.1 | ENST00000251413.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007904.hg.1 | ENST00000251413.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007904.hg.1 | ENST00000251413.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007904.hg.1 | ENST00000251413.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007904.hg.1 | ENST00000251413.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007904.hg.1 | ENST00000251413.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007904.hg.1 | ENST00000251413.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007904.hg.1 | ENST00000251413.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007904.hg.1 | ENST00000251413.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007904.hg.1 | ENST00000251413.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007904.hg.1 | ENST00000251413.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007904.hg.1 | ENST00000588056.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007904.hg.1 | ENST00000588056.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007904.hg.1 | ENST00000588056.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007904.hg.1 | ENST00000588056.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007904.hg.1 | ENST00000588056.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007904.hg.1 | ENST00000588056.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007904.hg.1 | ENST00000589613.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007904.hg.1 | ENST00000589613.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007904.hg.1 | ENST00000589613.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007904.hg.1 | ENST00000589613.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007904.hg.1 | ENST00000589688.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007904.hg.1 | ENST00000589688.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007904.hg.1 | ENST00000589688.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007904.hg.1 | ENST00000589688.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007904.hg.1 | ENST00000589688.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007904.hg.1 | ENST00000589688.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007904.hg.1 | ENST00000589688.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007904.hg.1 | ENST00000589688.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007904.hg.1 | ENST00000589688.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007904.hg.1 | ENST00000589688.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007904.hg.1 | ENST00000589688.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007904.hg.1 | ENST00000589688.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007904.hg.1 | ENST00000589688.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007904.hg.1 | ENST00000591509.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007904.hg.1 | ENST00000591509.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007904.hg.1 | ENST00000591509.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007904.hg.1 | ENST00000591509.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007904.hg.1 | ENST00000591509.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007904.hg.1 | ENST00000591509.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007904.hg.1 | ENST00000591509.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007904.hg.1 | ENST00000591509.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007904.hg.1 | ENST00000591509.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007904.hg.1 | ENST00000591509.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007904.hg.1 | NM_001070.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007904.hg.1 | NM_001070.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007904.hg.1 | NM_001070.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007904.hg.1 | NM_001070.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007904.hg.1 | NM_001070.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007904.hg.1 | NM_001070.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007904.hg.1 | NM_001070.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007904.hg.1 | NM_001070.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007904.hg.1 | NM_001070.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007904.hg.1 | NM_001070.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007904.hg.1 | NM_001070.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007904.hg.1 | NM_001070.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007904.hg.1 | NM_001070.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007904.hg.1 | NM_001070.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007904.hg.1 | NM_001070.4 | Refseq |
