Assay Details
| Gene Symbol: HCAR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006018.2 | NP_006009.2 | 1 | 655 | 83 | |
| GenBank mRNA | AK290930.1 | - | 1 | 536 | 83 | |
| AK313212.1 | - | 1 | 554 | 83 | ||
| BC038955.1 | - | 1 | 545 | 83 | ||
| BC047891.1 | - | 1 | 560 | 5750043 | 83 | |
| BI908162.1 | - | 1 | 547 | 83 | ||
| CU451507.1 | - | 1 | 83 | |||
| D10923.1 | - | 1 | 560 | 83 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29096579 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29096545 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08659943 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08659955 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29096553 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29096558 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08659959 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29096560 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08659963 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29096557 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29096565 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08659960 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08659967 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08659965 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16598850 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08659954 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29096554 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29096563 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29096561 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29096569 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08659968 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08659953 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16598829 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08659945 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08659946 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08659961 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08659962 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 875153 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P64721 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_23_P64721 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 11813 | - | - |
| Agilent Human Genome CGH Microarray 44B | A_14_P111297 | - | - |
| Agilent Human Genome CGH Microarray 44A | A_14_P111297 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 53644 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 27887 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P329924 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P64721 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_23_P64721 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | ERCC-00004_125 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08659957 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08659941 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29096566 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29096569 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29096553 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08659965 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08659953 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08659963 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08659959 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29096560 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08659946 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08659948 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08659960 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29096563 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08659940 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29096557 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08659943 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08659954 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29096554 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29096565 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08659945 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08659952 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29096561 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08659952 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08659965 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29096561 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08659959 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29096557 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08659954 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08659953 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08659948 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08659941 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08659946 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08659940 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29096553 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08659945 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08659960 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29096565 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08659943 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08659963 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29096563 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08659957 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29096566 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29096554 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29096569 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29096560 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_23_P64721 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P08659940 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P08659956 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P08659954 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P08659958 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P08659945 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P08659947 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P08659942 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P08659943 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P08659953 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P08659957 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P08659949 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P08659951 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P39844503 | - | - |
| Agilent Human Genome CGH Microarray 4x44K | A_14_P111297 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_23_P64721 | - | - |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 158931 | AF048977.1 | genbank_mrna_human |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 158931 | AF419855.1 | genbank_mrna_human |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 56281 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08659954 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08659958 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16598851 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08659964 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08659962 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08659956 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08659945 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08659960 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08659970 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08659968 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08659966 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16598853 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08659953 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08659951 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | NM_006018_at | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7967319 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7967323 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7967324 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 7967322 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 7967318 | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 205220_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 205220_at | - | - |
| Human Genome U133 Plus 2.0 Array | 205220_PM_at | - | - |
| Human Genome U133A 2.0 Array | 205220_at | - | - |
| Human Genome U133A 2.0 Array | 205220_PM_at | - | - |
| Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) | NM_006018_at | - | - |
| AffymetrixPathwork Diagnostics Pathchip | pw535605 | - | - |
| Human Genome U95 Set | 34951_at | - | - |
| Human Genome U95 Set | 34951_PM_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 205220_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 205220_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 205220_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 205220_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 205220_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 205220_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 205220_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 205220_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 205220_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 205220_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 205220_at3 | - | - |
| Human Genome U133 Set | 205220_PM_at | - | - |
| Human Genome U133 Set | 205220_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A048125 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A048125 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 134579 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_1677693 | BC063461.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1677693 | D10923.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1677693 | DA948030.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1677693 | BC038955.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1677693 | BC047891.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1791138 | - | - |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1782237 | - | - |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1677693 | BC063461.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1677693 | D10923.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1677693 | DA948030.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1677693 | BC038955.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1677693 | BC047891.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1677693 | BC063461.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1677693 | D10923.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1677693 | DA948030.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1677693 | BC038955.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1677693 | BC047891.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1677693 | BC063461.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1677693 | D10923.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1677693 | DA948030.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1677693 | BC038955.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1677693 | BC047891.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1677693 | BC063461.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1677693 | D10923.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1677693 | DA948030.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1677693 | BC038955.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1677693 | BC047891.1 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_5174460-S | D10923.1 | human_transcriptome |
| Illumina HumanRef-8 v2 Expression BeadChip | GI_22584-A | - | - |
| Illumina human-6 v1.0 expression beadchip | 0004760538 | - | - |
| Illumina humanRef-8 v1.0 expression beadchip | 0004760538 | - | - |
| Illumina human-6 v2.0 expression beadchip | ILMN_1791138 | - | - |
| Illumina human-6 v2.0 expression beadchip | ILMN_1677693 | BC063461.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1677693 | D10923.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1677693 | DA948030.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1677693 | BC038955.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1677693 | BC047891.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1782237 | - | - |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1677693 | BC063461.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1677693 | D10923.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1677693 | DA948030.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1677693 | BC038955.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1677693 | BC047891.1 | human_transcriptome |
| Illumina Sentrix Human-6 v2 Expression BeadChip | 5960360 | - | - |
| Illumina HumanRef-6 v2.0 expression beadchip | ILMN_22584 | - | - |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1677693 | BC063461.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1677693 | D10923.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1677693 | DA948030.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1677693 | BC038955.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1677693 | BC047891.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1677693 | BC063461.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1677693 | D10923.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1677693 | DA948030.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1677693 | BC038955.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1677693 | BC047891.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1791138 | - | - |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1782237 | - | - |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1791138 | - | - |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1677693 | BC063461.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1677693 | D10923.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1677693 | DA948030.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1677693 | BC038955.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1677693 | BC047891.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1782237 | - | - |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1677693 | BC063461.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1677693 | D10923.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1677693 | DA948030.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1677693 | BC038955.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1677693 | BC047891.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1677693 | BC063461.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1677693 | D10923.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1677693 | DA948030.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1677693 | BC038955.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1677693 | BC047891.1 | human_transcriptome |

HCAR3
Genomic Map