Assay Details
| Gene Symbol: PPIAL4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001143883.2 | NP_001137355.1 | 1 | 427 | 149 | |
| GenBank mRNA | AB084917.1 | - | 1 | 427 | 149 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01001163.hg.1
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20551022 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15120240 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 933138 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 509367 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 246115 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P203226 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P144499 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_32_P96272 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_1926 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_32_P61061 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_24_P264772 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_24_P144499 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_1925 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_24_P203226 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 30452 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 7703 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 14771 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 3615 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 14740 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 3193 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 6066 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P203226 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P144499 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P144499 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P203226 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P203226 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P144499 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_24_P144499 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_24_P203226 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20551022 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15120240 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20551022 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15120240 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_24_P264772 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_24_P144499 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_24_P203226 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_24_P264772 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_24_P203226 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_32_P96272 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_32_P61061 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_24_P144499 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | NM_178230_at | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7905052 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7919163 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7919164 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7919158 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7919437 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7919159 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7919410 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7919163 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7919164 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7905017 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7905017 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7905017 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7905052 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7919409 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7919164 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7905053 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7919411 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7919408 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7919158 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7919409 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7919410 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7919159 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7919408 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7904998 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7919165 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7919410 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7919409 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7919165 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7919158 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7919437 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7919437 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7919165 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7919159 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7904998 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7919411 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7919411 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7905053 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7919163 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7905052 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7904998 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 7919157 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 7919436 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 7905051 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 7919407 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 7905016 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 7919162 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 7904997 | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 217136_PM_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | PSR01017894.hg.1 | TC01001163.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01017894.hg.1 | ENST00000369222.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01017894.hg.1 | NM_001135789.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01017894.hg.1 | NM_001143883.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01017892.hg.1 | TC01001163.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01017892.hg.1 | ENST00000369222.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01017892.hg.1 | NM_001135789.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01017892.hg.1 | NM_001143883.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015660.hg.1 | ENST00000419275.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015660.hg.1 | ENST00000419275.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015660.hg.1 | ENST00000419275.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015660.hg.1 | ENST00000419275.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015660.hg.1 | ENST00000419275.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015660.hg.1 | NM_001123068.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015660.hg.1 | NM_001123068.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015660.hg.1 | NM_001123068.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009651.hg.1 | ENST00000577856.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009651.hg.1 | ENST00000577856.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009651.hg.1 | ENST00000577856.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009651.hg.1 | NM_001143883.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009651.hg.1 | NM_001143883.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009651.hg.1 | NM_001143883.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009651.hg.1 | NM_001143883.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015660.hg.1 | ENST00000419275.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015660.hg.1 | ENST00000419275.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015660.hg.1 | ENST00000419275.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015660.hg.1 | ENST00000419275.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015660.hg.1 | ENST00000419275.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015660.hg.1 | NM_001123068.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015660.hg.1 | NM_001123068.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015660.hg.1 | NM_001123068.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100169407.hg.1 | ENST00000577856.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100169407.hg.1 | NM_001143883.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100169407.hg.1 | ENST00000577856.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100169407.hg.1 | NM_001143883.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100169407.hg.1 | TC0100009651.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100169408.hg.1 | NM_001143883.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100169408.hg.1 | NM_001143883.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100169408.hg.1 | TC0100009651.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100009651.hg.1 | ENST00000577856.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009651.hg.1 | ENST00000577856.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009651.hg.1 | ENST00000577856.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009651.hg.1 | NM_001143883.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009651.hg.1 | NM_001143883.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009651.hg.1 | NM_001143883.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009651.hg.1 | NM_001143883.3 | Refseq |
| Human Genome U133 Plus 2.0 Array | 217136_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 217136_at | - | - |
| Human Genome U133 Plus 2.0 Array | 217136_at | - | - |
| Human Genome U133 Plus 2.0 Array | 217136_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 217136_at | - | - |
| Human Genome U133 Plus 2.0 Array | 217136_PM_at | - | - |
| Human Genome U133A 2.0 Array | 217136_PM_at | - | - |
| Human Genome U133A 2.0 Array | 217136_PM_at | - | - |
| Human Genome U133A 2.0 Array | 217136_PM_at | - | - |
| Human Genome U133A 2.0 Array | 217136_at | - | - |
| Human Genome U133A 2.0 Array | 217136_at | - | - |
| Human Genome U133A 2.0 Array | 217136_at | - | - |
| Human Genome U133 Set | 217136_PM_at | - | - |
| Human Genome U133 Set | 217136_at | - | - |
| Human Genome U133 Set | 217136_at | - | - |
| Human Genome U133 Set | 217136_at | - | - |
| Human Genome U133 Set | 217136_PM_at | - | - |
| Human Genome U133 Set | 217136_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A031070 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A050955 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A031070 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A051538 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A050955 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A051538 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A050955 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A050954 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A051538 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A031070 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A050954 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A031070 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A050954 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A051538 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A050955 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A051538 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A050955 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A051538 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A031070 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A050954 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A050955 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A031070 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 170108 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3241899 | - | - |
| HumanHT-12 v4 Expression BeadChip | ILMN_3236259 | NM_001135789.2 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3236259 | NM_001143883.2 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3236259 | NM_178230.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3236259 | AB084917.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1776260 | NM_001135789.2 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1776260 | NM_001143883.2 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1776260 | BC130378.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1776260 | AK123006.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1776260 | BC130376.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1712688 | - | - |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1705073 | AK123006.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3241899 | - | - |
| HumanHT-12 V4.0 expression beadchip | ILMN_3236259 | NM_001135789.2 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3236259 | NM_001143883.2 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3236259 | NM_178230.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3236259 | AB084917.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3236259 | NM_001135789.2 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3236259 | NM_001143883.2 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3236259 | NM_178230.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3236259 | AB084917.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3241899 | - | - |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3241899 | - | - |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3236259 | NM_001135789.2 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3236259 | NM_001143883.2 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3236259 | NM_178230.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3236259 | AB084917.1 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_30102943-S | NM_001144032.1 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_30102943-S | NM_001164262.1 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_30102943-S | NM_001135789.2 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_30102943-S | NM_001143883.2 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_30102943-S | NM_001164261.1 | human_transcriptome |
| Illumina HumanRef-8 v2 Expression BeadChip | GI_1203-S | - | - |
| Illumina human-6 v1.0 expression beadchip | 0002630746 | - | - |
| Illumina humanRef-8 v1.0 expression beadchip | 0002630746 | - | - |
| Illumina human-6 v2.0 expression beadchip | ILMN_1776260 | NM_001135789.2 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1776260 | NM_001143883.2 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1776260 | BC130378.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1776260 | AK123006.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1776260 | BC130376.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1705073 | AK123006.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1712688 | - | - |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1776260 | NM_001135789.2 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1776260 | NM_001143883.2 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1776260 | BC130378.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1776260 | AK123006.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1776260 | BC130376.1 | human_transcriptome |
| Illumina Sentrix Human-6 v2 Expression BeadChip | 5360255 | - | - |
| Illumina HumanRef-6 v2.0 expression beadchip | ILMN_1203 | - | - |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1776260 | NM_001135789.2 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1776260 | NM_001143883.2 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1776260 | BC130378.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1776260 | AK123006.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1776260 | BC130376.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1705073 | AK123006.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1699575 | - | - |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1699575 | - | - |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1776260 | NM_001135789.2 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1776260 | NM_001143883.2 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1776260 | BC130378.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1776260 | AK123006.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1776260 | BC130376.1 | human_transcriptome |

PPIAL4A
Genomic Map