Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009927.hg.1 |
ENST00000377602.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009927.hg.1 |
ENST00000377602.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009927.hg.1 |
ENST00000377602.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009927.hg.1 |
ENST00000377602.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009927.hg.1 |
ENST00000377602.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009927.hg.1 |
ENST00000377602.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009927.hg.1 |
ENST00000377602.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009927.hg.1 |
ENST00000377602.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009927.hg.1 |
ENST00000377602.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009927.hg.1 |
ENST00000377602.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009927.hg.1 |
ENST00000377602.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009927.hg.1 |
ENST00000377602.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009927.hg.1 |
ENST00000377602.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009927.hg.1 |
ENST00000440449.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009927.hg.1 |
ENST00000440449.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009927.hg.1 |
ENST00000440449.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009927.hg.1 |
ENST00000440449.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009927.hg.1 |
NM_001004470.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009927.hg.1 |
NM_001004470.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009927.hg.1 |
NM_001004470.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009927.hg.1 |
NM_001004470.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009927.hg.1 |
NM_001004470.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009927.hg.1 |
NM_001004470.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009927.hg.1 |
NM_001004470.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009927.hg.1 |
NM_001004470.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009927.hg.1 |
NM_001004470.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009927.hg.1 |
NM_001004470.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009927.hg.1 |
NM_001004470.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10013598.hg.1 |
ENST00000377602.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10013598.hg.1 |
ENST00000377610.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10013598.hg.1 |
NM_001004470.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10013598.hg.1 |
TC10001074.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR10013599.hg.1 |
ENST00000377602.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10013599.hg.1 |
ENST00000377610.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10013599.hg.1 |
NM_001004470.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10013599.hg.1 |
TC10001074.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1000167874.hg.1 |
ENST00000377602.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000167874.hg.1 |
NM_001004470.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000167874.hg.1 |
ENST00000377602.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000167874.hg.1 |
NM_001004470.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000167874.hg.1 |
TC1000009927.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1000009927.hg.1 |
ENST00000377602.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009927.hg.1 |
ENST00000377602.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009927.hg.1 |
ENST00000377602.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009927.hg.1 |
ENST00000377602.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009927.hg.1 |
ENST00000377602.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009927.hg.1 |
ENST00000377602.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009927.hg.1 |
ENST00000377602.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009927.hg.1 |
ENST00000377602.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009927.hg.1 |
ENST00000377602.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009927.hg.1 |
ENST00000377602.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009927.hg.1 |
ENST00000377602.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009927.hg.1 |
ENST00000377602.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009927.hg.1 |
ENST00000377602.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009927.hg.1 |
ENST00000440449.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009927.hg.1 |
ENST00000440449.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009927.hg.1 |
ENST00000440449.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009927.hg.1 |
ENST00000440449.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009927.hg.1 |
NM_001004470.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009927.hg.1 |
NM_001004470.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009927.hg.1 |
NM_001004470.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009927.hg.1 |
NM_001004470.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009927.hg.1 |
NM_001004470.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009927.hg.1 |
NM_001004470.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009927.hg.1 |
NM_001004470.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009927.hg.1 |
NM_001004470.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009927.hg.1 |
NM_001004470.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009927.hg.1 |
NM_001004470.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009927.hg.1 |
NM_001004470.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000167875.hg.1 |
ENST00000377602.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000167875.hg.1 |
ENST00000377602.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000167875.hg.1 |
TC1000009927.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000167873.hg.1 |
ENST00000377602.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000167873.hg.1 |
NM_001004470.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000167873.hg.1 |
ENST00000377602.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000167873.hg.1 |
NM_001004470.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000167873.hg.1 |
TC1000009927.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000167876.hg.1 |
TC1000009927.hg.1 |
Clariom D TC |