Assay Details
| Gene Symbol: OR5R1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004744.1 | NP_001004744.1 | 1 | 839 | 144 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11001776.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218847 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218843 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218813 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218835 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218792 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218841 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07809331 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218830 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218838 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218808 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218842 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218800 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16453359 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218818 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07809317 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218816 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218791 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07809342 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07809329 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218845 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07809314 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07809303 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218825 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218834 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07809315 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218829 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07809352 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218804 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218815 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218803 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07809300 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218827 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218819 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218788 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07809353 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07809335 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07809304 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07809349 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218812 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218820 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218796 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218801 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218836 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07809319 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07809345 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07809305 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218811 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218802 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28218848 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07809332 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3348594 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_0479 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_0480 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 1870 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3348594 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3348594 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218841 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218816 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07809342 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218808 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07809308 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218788 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07809349 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07809331 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218832 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218829 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218819 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218814 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218797 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07809332 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07809303 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218815 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07809335 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07809337 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07809329 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07809352 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218802 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07809319 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218804 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07809316 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07809305 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218800 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218818 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07809300 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218838 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218801 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218843 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218803 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218791 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16453359 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07809315 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218837 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218834 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218813 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07809317 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07809332 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218791 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218802 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218841 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218816 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07809335 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218819 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07809349 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07809331 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07809319 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218801 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07809337 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07809317 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218832 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218834 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218803 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07809315 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218837 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218843 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218808 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07809305 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07809329 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218804 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218829 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218814 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218813 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07809308 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07809300 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16453359 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218815 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218838 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218788 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07809342 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07809316 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218800 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07809352 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218818 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07809303 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28218797 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P07809295 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P07809335 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P07809296 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P07809325 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P07809328 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P07809345 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P07809317 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P07809333 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P07809309 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P07809300 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P07809323 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P07809312 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P07809307 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P07809308 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P07809313 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P07809305 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P07809298 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P07809303 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P07809331 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P07809326 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P07809316 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P07809321 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P07809338 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P07809318 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P07809324 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P07809337 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P07809293 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P07809348 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P07809343 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P07809301 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07809307 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16453363 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07809316 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07809318 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07809301 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07809328 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07809358 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07809309 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07809313 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07809338 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07809305 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07809351 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07809349 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07809355 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07809308 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07809323 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07809300 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07809345 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16453360 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07809333 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07809353 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07809343 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07809321 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07809335 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07809312 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07809303 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07809324 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07809331 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07809357 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07809347 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07809317 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07809326 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07809337 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | NM_001004744_at | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7948130 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 7948129 | - | - |
| Affymetrix Human Genome U219 Array | 11742545_PM_at | - | - |
| Affymetrix Human Genome U219 Array | 11742545_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010855.hg.1 | ENST00000312253.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010855.hg.1 | ENST00000623216.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010855.hg.1 | ENST00000623286.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010855.hg.1 | NM_001004744.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11020678.hg.1 | ENST00000312253.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11020678.hg.1 | NM_001004744.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11020678.hg.1 | TC11001776.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1100010855.hg.1 | ENST00000312253.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010855.hg.1 | ENST00000623216.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010855.hg.1 | ENST00000623286.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010855.hg.1 | NM_001004744.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100182113.hg.1 | ENST00000312253.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100182113.hg.1 | ENST00000623216.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100182113.hg.1 | ENST00000623286.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100182113.hg.1 | NM_001004744.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100182113.hg.1 | ENST00000312253.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100182113.hg.1 | ENST00000623216.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100182113.hg.1 | ENST00000623286.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100182113.hg.1 | NM_001004744.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100182113.hg.1 | TC1100010855.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A045166 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A045166 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_1655449 | NM_001004744.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1655449 | NM_001004744.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1655449 | NM_001004744.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1655449 | NM_001004744.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1655449 | NM_001004744.1 | human_transcriptome |
| Illumina HumanRef-8 v2 Expression BeadChip | GI_5401-S | - | - |
| Illumina human-6 v2.0 expression beadchip | ILMN_1655449 | NM_001004744.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1655449 | NM_001004744.1 | human_transcriptome |
| Illumina Sentrix Human-6 v2 Expression BeadChip | ILMN_5401 | - | - |
| Illumina HumanRef-6 v2.0 expression beadchip | ILMN_5401 | - | - |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1655449 | NM_001004744.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1655449 | NM_001004744.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1655449 | NM_001004744.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1655449 | NM_001004744.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1655449 | NM_001004744.1 | human_transcriptome |

OR5R1
Genomic Map