Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010015.hg.1 |
ENST00000625487.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010015.hg.1 |
ENST00000625487.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006946.hg.1 |
ENST00000389078.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006946.hg.1 |
ENST00000389078.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006946.hg.1 |
ENST00000389078.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006946.hg.1 |
ENST00000389078.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006946.hg.1 |
ENST00000389078.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006946.hg.1 |
ENST00000501744.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006946.hg.1 |
ENST00000501744.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006946.hg.1 |
ENST00000501744.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006946.hg.1 |
ENST00000501744.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006946.hg.1 |
ENST00000501744.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006946.hg.1 |
ENST00000544848.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006946.hg.1 |
ENST00000544848.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006946.hg.1 |
ENST00000544848.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006946.hg.1 |
ENST00000544848.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006946.hg.1 |
ENST00000544848.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006946.hg.1 |
ENST00000544848.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006946.hg.1 |
ENST00000544848.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006946.hg.1 |
NM_177925.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006946.hg.1 |
NM_177925.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006946.hg.1 |
NM_177925.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006946.hg.1 |
NM_177925.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006946.hg.1 |
NM_177925.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006946.hg.1 |
NM_177925.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006946.hg.1 |
NM_177925.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006946.hg.1 |
NM_177925.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006946.hg.1 |
NM_177925.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006946.hg.1 |
NM_177925.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006946.hg.1 |
NM_177925.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006946.hg.1 |
NR_027716.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006946.hg.1 |
NR_027716.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006946.hg.1 |
NR_027716.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006946.hg.1 |
NR_027716.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006946.hg.1 |
NR_027716.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006946.hg.1 |
NR_027716.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006946.hg.1 |
NR_027716.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006946.hg.1 |
NR_027716.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006946.hg.1 |
NR_027716.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12002458.hg.1 |
ENST00000544848.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12002458.hg.1 |
NM_177925.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12002458.hg.1 |
NR_027716.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12002458.hg.1 |
TC12000197.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1200148549.hg.1 |
ENST00000501744.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200148549.hg.1 |
ENST00000544848.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200148549.hg.1 |
NM_177925.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200148549.hg.1 |
NR_027716.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200148549.hg.1 |
ENST00000501744.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200148549.hg.1 |
ENST00000544848.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200148549.hg.1 |
NM_177925.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200148549.hg.1 |
NR_027716.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200148549.hg.1 |
TC1200006946.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200148548.hg.1 |
ENST00000389078.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200148548.hg.1 |
ENST00000501744.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200148548.hg.1 |
ENST00000544848.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200148548.hg.1 |
NM_177925.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200148548.hg.1 |
NR_027716.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200148548.hg.1 |
ENST00000389078.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200148548.hg.1 |
ENST00000501744.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200148548.hg.1 |
ENST00000544848.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200148548.hg.1 |
NM_177925.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200148548.hg.1 |
NR_027716.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200148548.hg.1 |
TC1200006946.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1200006946.hg.1 |
ENST00000389078.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006946.hg.1 |
ENST00000389078.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006946.hg.1 |
ENST00000389078.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006946.hg.1 |
ENST00000389078.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006946.hg.1 |
ENST00000389078.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006946.hg.1 |
ENST00000501744.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006946.hg.1 |
ENST00000501744.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006946.hg.1 |
ENST00000501744.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006946.hg.1 |
ENST00000501744.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006946.hg.1 |
ENST00000501744.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006946.hg.1 |
ENST00000544848.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006946.hg.1 |
ENST00000544848.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006946.hg.1 |
ENST00000544848.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006946.hg.1 |
ENST00000544848.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006946.hg.1 |
ENST00000544848.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006946.hg.1 |
ENST00000544848.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006946.hg.1 |
ENST00000544848.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006946.hg.1 |
NM_177925.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006946.hg.1 |
NM_177925.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006946.hg.1 |
NM_177925.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006946.hg.1 |
NM_177925.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006946.hg.1 |
NM_177925.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006946.hg.1 |
NM_177925.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006946.hg.1 |
NM_177925.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006946.hg.1 |
NM_177925.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006946.hg.1 |
NM_177925.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006946.hg.1 |
NM_177925.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006946.hg.1 |
NM_177925.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006946.hg.1 |
NR_027716.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006946.hg.1 |
NR_027716.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006946.hg.1 |
NR_027716.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006946.hg.1 |
NR_027716.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006946.hg.1 |
NR_027716.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006946.hg.1 |
NR_027716.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006946.hg.1 |
NR_027716.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006946.hg.1 |
NR_027716.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006946.hg.1 |
NR_027716.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010015.hg.1 |
ENST00000625487.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010015.hg.1 |
ENST00000625487.1 |
Ensembl |