Assay Details
| Gene Symbol: LINC00167 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_024233.1 | 1 | 2509 | 151 | ||
| GenBank mRNA | AK055956.1 | - | 2 | 2361 | 151 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11001193.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 50140 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P359030 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_24_P359030 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 6190 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 3986 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 12638 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P359030 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_24_P359030 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08118360 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28546166 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28546197 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28546180 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08118357 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28546175 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28546224 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28546174 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08118302 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28546219 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28546163 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28546165 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28546164 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28546170 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28546184 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28546179 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08118337 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28546173 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28546150 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28546187 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28546194 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28546185 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28546171 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28546167 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08118364 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08118317 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08118308 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08118365 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28546153 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28546225 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28546151 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28546228 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08118359 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28546182 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08118314 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08118331 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08118325 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08118305 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28546227 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28546158 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08118361 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28546181 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08118317 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08118359 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28546150 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08118314 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28546180 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08118364 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28546164 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28546185 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28546167 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28546165 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28546219 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28546181 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08118365 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08118305 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28546184 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08118361 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28546227 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08118331 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28546197 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28546175 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08118360 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28546153 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28546194 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28546173 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28546166 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08118337 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08118325 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28546158 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08118308 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08118302 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28546225 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28546228 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28546224 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28546170 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28546171 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28546151 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28546182 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28546174 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08118357 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28546163 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28546179 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28546187 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_24_P359030 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_24_P144104 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_24_P359030 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_24_P144104 | - | - |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 127312 | AK024282.1 | genbank_mrna_human |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 127312 | BC030637.1 | genbank_mrna_human |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 127312 | NM_024893.1 | refseq_mrna_human_NM |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 56958 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P19417379 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P33084194 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08118334 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08118324 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08118331 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08118314 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08118298 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08118327 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16516660 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08118363 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08118319 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16516665 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08118306 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08118308 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08118316 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08118366 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08118336 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08118359 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08118358 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08118361 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08118364 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08118303 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08118338 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16516655 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08118312 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08118299 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16516662 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16516658 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08118330 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08118370 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08118310 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16516670 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08118322 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P19417379 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | 7945180 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7945178 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7945181 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7945179 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 7945177 | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 1562413_PM_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | PSR11014989.hg.1 | NR_024233.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11014989.hg.1 | TC11001193.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR11014988.hg.1 | ENST00000530583.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11014988.hg.1 | NR_024233.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11014988.hg.1 | TC11001193.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR11014987.hg.1 | NR_024233.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11014987.hg.1 | TC11001193.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1100171516.hg.1 | ENST00000530583.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100171516.hg.1 | NR_024233.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100171516.hg.1 | ENST00000530583.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100171516.hg.1 | NR_024233.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100171516.hg.1 | TC1100009517.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100171517.hg.1 | ENST00000530583.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100171517.hg.1 | NR_024233.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100171517.hg.1 | ENST00000530583.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100171517.hg.1 | NR_024233.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100171517.hg.1 | TC1100009517.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100171518.hg.1 | NR_024233.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100171518.hg.1 | NR_024233.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100171518.hg.1 | TC1100009517.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100171519.hg.1 | NR_024233.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100171519.hg.1 | NR_024233.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100171519.hg.1 | TC1100009517.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100171515.hg.1 | NR_024233.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100171515.hg.1 | NR_024233.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100171515.hg.1 | TC1100009517.hg.1 | Clariom D TC |
| Human Genome U133 Plus 2.0 Array | 1562413_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1562413_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562413_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562413_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562413_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562413_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562413_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562413_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562413_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562413_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562413_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562413_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562413_at9 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A047142 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A047142 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3244888 | AK055956.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3244888 | NR_024233.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1789804 | AK055956.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1789804 | NR_024233.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3244888 | AK055956.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3244888 | NR_024233.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3244888 | AK055956.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3244888 | NR_024233.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1789804 | AK055956.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1789804 | NR_024233.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3244888 | AK055956.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3244888 | NR_024233.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1789804 | AK055956.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1789804 | NR_024233.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1789804 | AK055956.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1789804 | NR_024233.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1789804 | AK055956.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1789804 | NR_024233.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1789804 | AK055956.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1789804 | NR_024233.1 | human_transcriptome |

LINC00167
Genomic Map