Assay Details
Gene Symbol: CHST11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001173982.1 | NP_001167453.1 | 3 | 1761 | 102 | |
NM_018413.5 | NP_060883.1 | 3 | 1776 | 102 | ||
XM_017019369.1 | XP_016874858.1 | 5 | 1347 | 102 | ||
GenBank mRNA | AF239820.1 | - | 3 | 1427 | 102 | |
BC013315.1 | - | 4 | 1573 | 4042813 | 102 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC12000812.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
chondrocyte development
respiratory gaseous exchange
post-embryonic development
carbohydrate biosynthetic process
chondroitin sulfate biosynthetic process
negative regulation of transforming growth factor beta receptor signaling pathway
polysaccharide localization
post-anal tail morphogenesis
regulation of cell proliferation
embryonic digit morphogenesis
negative regulation of apoptotic process
developmental growth
embryonic viscerocranium morphogenesis
N-acetylgalactosamine 4-O-sulfotransferase activity
chondroitin 4-sulfotransferase activity
N-acetylgalactosamine 4-sulfate 6-O-sulfotransferase activity
chondrocyte development
respiratory gaseous exchange
post-embryonic development
carbohydrate biosynthetic process
chondroitin sulfate biosynthetic process
negative regulation of transforming growth factor beta receptor signaling pathway
polysaccharide localization
post-anal tail morphogenesis
regulation of cell proliferation
embryonic digit morphogenesis
negative regulation of apoptotic process
developmental growth
embryonic viscerocranium morphogenesis
N-acetylgalactosamine 4-O-sulfotransferase activity
chondroitin 4-sulfotransferase activity
N-acetylgalactosamine 4-sulfate 6-O-sulfotransferase activity
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | ENST00000303694.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | ENST00000303694.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | ENST00000303694.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | ENST00000303694.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | ENST00000303694.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | ENST00000303694.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | ENST00000303694.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | ENST00000303694.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | ENST00000303694.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | ENST00000303694.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | ENST00000546689.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | ENST00000546689.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | ENST00000546689.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | ENST00000546689.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | ENST00000546689.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | ENST00000546689.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | ENST00000547956.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | ENST00000547956.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | ENST00000547956.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | ENST00000547956.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | ENST00000547956.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | ENST00000547956.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | ENST00000547956.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | ENST00000549016.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | ENST00000549016.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | ENST00000549016.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | ENST00000549260.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | ENST00000549260.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | ENST00000549260.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | ENST00000549260.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | ENST00000549260.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | ENST00000549260.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | ENST00000549260.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | ENST00000549260.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | ENST00000549260.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | ENST00000549260.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | ENST00000550711.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | ENST00000550711.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | NM_001173982.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | NM_001173982.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | NM_001173982.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | NM_001173982.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | NM_001173982.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | NM_001173982.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | NM_001173982.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | NM_001173982.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | NM_001173982.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | NM_001173982.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | NM_001173982.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | NM_018413.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | NM_018413.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | NM_018413.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | NM_018413.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | NM_018413.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | NM_018413.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | NM_018413.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | NM_018413.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | NM_018413.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | NM_018413.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | NM_018413.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12010357.hg.1 | ENST00000303694.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12010357.hg.1 | ENST00000549260.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12010357.hg.1 | NM_001173982.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12010357.hg.1 | NM_018413.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12010357.hg.1 | TC12000812.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1200164166.hg.1 | ENST00000303694.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200164166.hg.1 | ENST00000549260.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200164166.hg.1 | NM_001173982.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200164166.hg.1 | NM_018413.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200164166.hg.1 | ENST00000303694.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200164166.hg.1 | ENST00000549260.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200164166.hg.1 | NM_001173982.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200164166.hg.1 | NM_018413.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200164166.hg.1 | TC1200008683.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | ENST00000303694.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | ENST00000303694.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | ENST00000303694.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | ENST00000303694.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | ENST00000303694.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | ENST00000303694.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | ENST00000303694.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | ENST00000303694.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | ENST00000303694.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | ENST00000303694.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | ENST00000546689.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | ENST00000546689.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | ENST00000546689.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | ENST00000546689.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | ENST00000546689.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | ENST00000546689.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | ENST00000547956.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | ENST00000547956.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | ENST00000547956.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | ENST00000547956.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | ENST00000547956.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | ENST00000547956.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | ENST00000547956.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | ENST00000549016.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | ENST00000549016.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | ENST00000549016.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | ENST00000549260.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | ENST00000549260.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | ENST00000549260.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | ENST00000549260.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | ENST00000549260.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | ENST00000549260.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | ENST00000549260.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | ENST00000549260.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | ENST00000549260.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | ENST00000549260.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | ENST00000550711.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | ENST00000550711.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | NM_001173982.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | NM_001173982.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | NM_001173982.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | NM_001173982.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | NM_001173982.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | NM_001173982.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | NM_001173982.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | NM_001173982.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | NM_001173982.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | NM_001173982.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | NM_001173982.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | NM_018413.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | NM_018413.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | NM_018413.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | NM_018413.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | NM_018413.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | NM_018413.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | NM_018413.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | NM_018413.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | NM_018413.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | NM_018413.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | NM_018413.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200164167.hg.1 | ENST00000303694.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200164167.hg.1 | ENST00000549260.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200164167.hg.1 | NM_001173982.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200164167.hg.1 | NM_018413.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200164167.hg.1 | ENST00000303694.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200164167.hg.1 | ENST00000549260.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200164167.hg.1 | NM_001173982.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200164167.hg.1 | NM_018413.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200164167.hg.1 | TC1200008683.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1200164165.hg.1 | ENST00000303694.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200164165.hg.1 | ENST00000549260.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200164165.hg.1 | NM_001173982.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200164165.hg.1 | NM_018413.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200164165.hg.1 | ENST00000303694.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200164165.hg.1 | ENST00000549260.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200164165.hg.1 | NM_001173982.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200164165.hg.1 | NM_018413.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200164165.hg.1 | TC1200008683.hg.1 | Clariom D TC |
