Assay Details
Gene Symbol: LOC728743 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK022764.1 | - | 1 - 2 | 273 | 105 | |
AK022851.1 | - | 1 - 2 | 211 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0700013490.hg.1 | ENST00000464630.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013490.hg.1 | ENST00000464630.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013490.hg.1 | ENST00000464630.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013490.hg.1 | ENST00000464630.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013490.hg.1 | ENST00000474836.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013490.hg.1 | ENST00000474836.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013490.hg.1 | ENST00000474836.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013490.hg.1 | ENST00000474836.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013490.hg.1 | ENST00000474836.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013490.hg.1 | ENST00000476489.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013490.hg.1 | ENST00000476489.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013490.hg.1 | ENST00000476489.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013490.hg.1 | ENST00000476489.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013490.hg.1 | ENST00000476489.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013490.hg.1 | ENST00000476489.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013490.hg.1 | ENST00000476489.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013490.hg.1 | ENST00000476489.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013490.hg.1 | ENST00000480389.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013490.hg.1 | ENST00000480389.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013490.hg.1 | ENST00000480389.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013490.hg.1 | ENST00000481877.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013490.hg.1 | ENST00000481877.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013490.hg.1 | ENST00000481877.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013490.hg.1 | ENST00000481877.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013490.hg.1 | ENST00000481877.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013490.hg.1 | ENST00000491664.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013490.hg.1 | ENST00000491664.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013490.hg.1 | ENST00000491664.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013490.hg.1 | ENST00000491664.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013490.hg.1 | ENST00000491664.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013490.hg.1 | ENST00000498682.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013490.hg.1 | ENST00000498682.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013490.hg.1 | ENST00000498682.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013490.hg.1 | ENST00000498682.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013490.hg.1 | ENST00000498682.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013490.hg.1 | ENST00000498682.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013490.hg.1 | ENST00000498682.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013490.hg.1 | NR_027237.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013490.hg.1 | NR_027237.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013490.hg.1 | NR_027237.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013490.hg.1 | NR_027237.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013490.hg.1 | NR_027237.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013490.hg.1 | NR_027237.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013490.hg.1 | NR_027237.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013490.hg.1 | NR_027237.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013490.hg.1 | NR_027237.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013490.hg.1 | NR_027237.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013490.hg.1 | NR_027237.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013490.hg.1 | NR_027237.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013490.hg.1 | NR_027237.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013490.hg.1 | ENST00000464630.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013490.hg.1 | ENST00000464630.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013490.hg.1 | ENST00000464630.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013490.hg.1 | ENST00000464630.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013490.hg.1 | ENST00000474836.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013490.hg.1 | ENST00000474836.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013490.hg.1 | ENST00000474836.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013490.hg.1 | ENST00000474836.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013490.hg.1 | ENST00000474836.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013490.hg.1 | ENST00000476489.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013490.hg.1 | ENST00000476489.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013490.hg.1 | ENST00000476489.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013490.hg.1 | ENST00000476489.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013490.hg.1 | ENST00000476489.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013490.hg.1 | ENST00000476489.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013490.hg.1 | ENST00000476489.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013490.hg.1 | ENST00000476489.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013490.hg.1 | ENST00000480389.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013490.hg.1 | ENST00000480389.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013490.hg.1 | ENST00000480389.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013490.hg.1 | ENST00000481877.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013490.hg.1 | ENST00000481877.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013490.hg.1 | ENST00000481877.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013490.hg.1 | ENST00000481877.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013490.hg.1 | ENST00000481877.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013490.hg.1 | ENST00000491664.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013490.hg.1 | ENST00000491664.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013490.hg.1 | ENST00000491664.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013490.hg.1 | ENST00000491664.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013490.hg.1 | ENST00000491664.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013490.hg.1 | ENST00000498682.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013490.hg.1 | ENST00000498682.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013490.hg.1 | ENST00000498682.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013490.hg.1 | ENST00000498682.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013490.hg.1 | ENST00000498682.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013490.hg.1 | ENST00000498682.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013490.hg.1 | ENST00000498682.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013490.hg.1 | NR_027237.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013490.hg.1 | NR_027237.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013490.hg.1 | NR_027237.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013490.hg.1 | NR_027237.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013490.hg.1 | NR_027237.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013490.hg.1 | NR_027237.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013490.hg.1 | NR_027237.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013490.hg.1 | NR_027237.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013490.hg.1 | NR_027237.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013490.hg.1 | NR_027237.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013490.hg.1 | NR_027237.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013490.hg.1 | NR_027237.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013490.hg.1 | NR_027237.1 | Refseq |
