Assay Details
| Gene Symbol: HRNR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009931.2 | NP_001009931.1 | 2 - 3 | 217 | 103 | |
| GenBank mRNA | AY396741.1 | - | 2 - 3 | 217 | 103 | |
More Information
Set Membership: |
|

HRNR
Genomic Map