Assay Details
| Gene Symbol: PLPP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005269592.2 | XP_005269649.1 | 6 | 10754 | 126 | |
| XM_006717685.3 | XP_006717748.1 | 7 | 10594 | 126 | ||
| XM_011539444.2 | XP_011537746.1 | 8 | 10652 | 126 | ||
| XM_017015821.1 | XP_016871310.1 | 4 | 10335 | 126 | ||
| XM_017015822.1 | XP_016871311.1 | 4 | 10302 | 126 | ||
| Gene Symbol: LINC01561 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_103717.1 | 1 | 1311 | 126 | ||
| GenBank mRNA | AK094721.1 | - | 1 | 1054 | 126 | |
| BC108662.1 | - | 1 | 1314 | 3922405 | 126 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27917117 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07526459 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27917112 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27917115 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07526480 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27917126 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27917113 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27917118 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07526473 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07526453 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07526444 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07526452 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27917120 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27917102 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07526445 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07526491 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27917146 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27917096 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27917108 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27917098 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27917140 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07526487 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07526498 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07526503 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07526486 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07526479 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27917121 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07526489 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07526450 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07526501 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07526483 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07526449 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27917133 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27917114 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07526461 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27917135 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27917142 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27917107 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07526493 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07526457 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27917104 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07526458 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07526496 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07526481 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27917127 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07526474 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 3335 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3259929 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | 7930995 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 7930994 | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 236855_PM_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012343.hg.1 | ENST00000521923.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012343.hg.1 | ENST00000521923.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012343.hg.1 | ENST00000521923.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012343.hg.1 | ENST00000521923.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012343.hg.1 | ENST00000521923.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009106.hg.1 | ENST00000369071.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009106.hg.1 | ENST00000369071.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009106.hg.1 | ENST00000369071.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009106.hg.1 | ENST00000623529.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009106.hg.1 | ENST00000623529.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009106.hg.1 | ENST00000623529.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009106.hg.1 | NR_103717.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009106.hg.1 | NR_103717.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009106.hg.1 | NR_103717.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009106.hg.1 | ENST00000369071.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009106.hg.1 | ENST00000369071.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009106.hg.1 | ENST00000369071.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009106.hg.1 | ENST00000623529.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009106.hg.1 | ENST00000623529.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009106.hg.1 | ENST00000623529.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009106.hg.1 | NR_103717.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009106.hg.1 | NR_103717.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009106.hg.1 | NR_103717.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000162317.hg.1 | ENST00000369071.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000162317.hg.1 | ENST00000623529.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000162317.hg.1 | NR_103717.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000162317.hg.1 | ENST00000369071.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000162317.hg.1 | ENST00000623529.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000162317.hg.1 | NR_103717.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000162317.hg.1 | TC1000009106.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000162318.hg.1 | ENST00000369071.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000162318.hg.1 | ENST00000623529.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000162318.hg.1 | NR_103717.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000162318.hg.1 | ENST00000369071.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000162318.hg.1 | ENST00000623529.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000162318.hg.1 | NR_103717.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000162318.hg.1 | TC1000009106.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000162319.hg.1 | ENST00000369071.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000162319.hg.1 | ENST00000623529.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000162319.hg.1 | NR_103717.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000162319.hg.1 | ENST00000369071.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000162319.hg.1 | ENST00000623529.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000162319.hg.1 | NR_103717.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000162319.hg.1 | TC1000009106.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0800012343.hg.1 | ENST00000521923.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012343.hg.1 | ENST00000521923.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012343.hg.1 | ENST00000521923.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012343.hg.1 | ENST00000521923.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012343.hg.1 | ENST00000521923.5 | Ensembl |
| Human Genome U133 Plus 2.0 Array | 236855_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 236855_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 236855_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 236855_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 236855_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 236855_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 236855_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 236855_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 236855_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 236855_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 236855_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 236855_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 236855_at2 | - | - |
| Human Genome U95 Set | 63668_PM_at | - | - |
| Human Genome U95 Set | 63668_at | - | - |
| Human Genome U133 Set | 236855_at | - | - |
| Human Genome U133 Set | 236855_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3307060 | BC108662.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3307060 | AK094721.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1665207 | BC108662.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1665207 | AK094721.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1665207 | BC108662.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1665207 | AK094721.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1665207 | BC108662.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1665207 | AK094721.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1665207 | BC108662.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1665207 | AK094721.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1665207 | BC108662.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1665207 | AK094721.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1665207 | BC108662.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1665207 | AK094721.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1665207 | BC108662.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1665207 | AK094721.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1665207 | BC108662.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1665207 | AK094721.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1665207 | BC108662.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1665207 | AK094721.1 | human_transcriptome |

PLPP4
Genomic Map