Assay Details
| Gene Symbol: RGPD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024457.3 | NP_001019628.3 | 1 - 2 | 57 | 146 | |
| XM_017004104.1 | XP_016859593.1 | 1 - 2 | 373 | 146 | ||
| GenBank mRNA | CR749330.1 | - | 1 - 2 | 57 | 146 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02002064.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR02033161.hg.1 | TC02002064.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02033161.hg.1 | ENST00000420840.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02033161.hg.1 | NM_001024457.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013351.hg.1 | ENST00000398146.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013351.hg.1 | ENST00000398146.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013351.hg.1 | ENST00000398146.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013351.hg.1 | ENST00000398146.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013351.hg.1 | ENST00000398146.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013351.hg.1 | ENST00000398146.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013351.hg.1 | ENST00000398146.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013351.hg.1 | ENST00000398146.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013351.hg.1 | ENST00000398146.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013351.hg.1 | ENST00000398146.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013351.hg.1 | ENST00000398146.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013351.hg.1 | ENST00000398146.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013351.hg.1 | ENST00000398146.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013351.hg.1 | ENST00000398146.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013351.hg.1 | ENST00000398146.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013351.hg.1 | ENST00000398146.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013351.hg.1 | ENST00000398146.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013351.hg.1 | ENST00000398146.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013351.hg.1 | ENST00000398146.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013351.hg.1 | ENST00000398146.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013351.hg.1 | ENST00000398146.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013351.hg.1 | ENST00000398146.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013351.hg.1 | ENST00000398146.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013351.hg.1 | ENST00000398146.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013351.hg.1 | ENST00000398146.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013351.hg.1 | ENST00000398146.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013351.hg.1 | ENST00000398146.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013351.hg.1 | ENST00000398146.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013351.hg.1 | ENST00000494592.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013351.hg.1 | ENST00000494592.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013351.hg.1 | ENST00000617314.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013351.hg.1 | ENST00000617314.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013351.hg.1 | ENST00000617314.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013351.hg.1 | ENST00000617314.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013351.hg.1 | ENST00000617314.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013351.hg.1 | ENST00000617314.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013351.hg.1 | ENST00000617314.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013351.hg.1 | NM_001024457.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013351.hg.1 | NM_001024457.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013351.hg.1 | NM_001024457.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013351.hg.1 | NM_001024457.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013351.hg.1 | NM_001024457.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013351.hg.1 | NM_001024457.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013351.hg.1 | NM_001024457.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013351.hg.1 | NM_001024457.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013351.hg.1 | NM_001024457.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013351.hg.1 | NM_001024457.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013351.hg.1 | NM_001024457.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013351.hg.1 | NM_001024457.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013351.hg.1 | NM_001024457.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013351.hg.1 | NM_001024457.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013351.hg.1 | NM_001024457.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013351.hg.1 | NM_001024457.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013351.hg.1 | NM_001024457.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013351.hg.1 | NM_001024457.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013351.hg.1 | NM_001024457.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013351.hg.1 | NM_001024457.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013351.hg.1 | NM_001024457.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013351.hg.1 | NM_001024457.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013351.hg.1 | NM_001024457.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013351.hg.1 | NM_001024457.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013351.hg.1 | NM_001024457.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013351.hg.1 | NM_001024457.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02033157.hg.1 | TC02002064.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02033157.hg.1 | ENST00000327544.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02033157.hg.1 | ENST00000398146.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02033157.hg.1 | ENST00000420840.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02033157.hg.1 | NM_001024457.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02033157.hg.1 | NM_001078170.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200197566.hg.1 | ENST00000398146.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200197566.hg.1 | NM_001024457.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200197566.hg.1 | ENST00000398146.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200197566.hg.1 | NM_001024457.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200197566.hg.1 | TC0200013351.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200197572.hg.1 | ENST00000617314.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200197572.hg.1 | NM_001024457.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200197572.hg.1 | ENST00000617314.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200197572.hg.1 | NM_001024457.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200197572.hg.1 | TC0200013351.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200013351.hg.1 | ENST00000398146.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013351.hg.1 | ENST00000398146.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013351.hg.1 | ENST00000398146.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013351.hg.1 | ENST00000398146.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013351.hg.1 | ENST00000398146.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013351.hg.1 | ENST00000398146.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013351.hg.1 | ENST00000398146.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013351.hg.1 | ENST00000398146.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013351.hg.1 | ENST00000398146.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013351.hg.1 | ENST00000398146.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013351.hg.1 | ENST00000398146.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013351.hg.1 | ENST00000398146.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013351.hg.1 | ENST00000398146.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013351.hg.1 | ENST00000398146.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013351.hg.1 | ENST00000398146.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013351.hg.1 | ENST00000398146.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013351.hg.1 | ENST00000398146.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013351.hg.1 | ENST00000398146.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013351.hg.1 | ENST00000398146.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013351.hg.1 | ENST00000398146.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013351.hg.1 | ENST00000398146.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013351.hg.1 | ENST00000398146.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013351.hg.1 | ENST00000398146.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013351.hg.1 | ENST00000398146.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013351.hg.1 | ENST00000398146.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013351.hg.1 | ENST00000398146.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013351.hg.1 | ENST00000398146.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013351.hg.1 | ENST00000398146.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013351.hg.1 | ENST00000494592.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013351.hg.1 | ENST00000494592.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013351.hg.1 | ENST00000617314.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013351.hg.1 | ENST00000617314.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013351.hg.1 | ENST00000617314.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013351.hg.1 | ENST00000617314.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013351.hg.1 | ENST00000617314.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013351.hg.1 | ENST00000617314.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013351.hg.1 | ENST00000617314.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013351.hg.1 | NM_001024457.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013351.hg.1 | NM_001024457.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013351.hg.1 | NM_001024457.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013351.hg.1 | NM_001024457.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013351.hg.1 | NM_001024457.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013351.hg.1 | NM_001024457.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013351.hg.1 | NM_001024457.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013351.hg.1 | NM_001024457.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013351.hg.1 | NM_001024457.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013351.hg.1 | NM_001024457.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013351.hg.1 | NM_001024457.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013351.hg.1 | NM_001024457.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013351.hg.1 | NM_001024457.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013351.hg.1 | NM_001024457.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013351.hg.1 | NM_001024457.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013351.hg.1 | NM_001024457.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013351.hg.1 | NM_001024457.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013351.hg.1 | NM_001024457.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013351.hg.1 | NM_001024457.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013351.hg.1 | NM_001024457.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013351.hg.1 | NM_001024457.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013351.hg.1 | NM_001024457.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013351.hg.1 | NM_001024457.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013351.hg.1 | NM_001024457.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013351.hg.1 | NM_001024457.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013351.hg.1 | NM_001024457.3 | Refseq |

RGPD1
Genomic Map