Assay Details
| Gene Symbol: PPIP5K1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF502589.1 | - | 27 - 28 | 3129 | 59 | |
| AF543190.1 | - | 27 - 28 | 3079 | 59 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PPIP5K1| Gene Symbol: PPIP5K1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF502589.1 | - | 27 - 28 | 3129 | 59 | |
| AF543190.1 | - | 27 - 28 | 3079 | 59 | ||
Set Membership: |
|