Assay Details
Gene Symbol: C7orf66 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024607.2 | NP_001019778.1 | 1 - 2 | 155 | 76 | |
GenBank mRNA | AI126292.1 | - | 1 - 2 | 76 | ||
BG211856.1 | - | 1 - 2 | 156 | 76 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC07001753.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25937811 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25937763 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25937800 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25937825 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25937799 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P05680934 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25937776 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25937812 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25937771 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25937792 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25937769 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P05680969 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P05680940 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25937819 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25937764 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25937788 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25937803 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25937786 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25937778 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25937796 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P05680950 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25937808 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25937823 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P05680941 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P05680978 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P05680976 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25937782 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25937793 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P05680925 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25937787 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P05680970 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25937822 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25937818 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25937821 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P05680937 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25937775 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P05680964 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P05680974 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P05680954 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P05680986 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25937813 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25937795 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25937765 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P05680955 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25937804 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25937805 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P05680935 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25937777 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25937790 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25937802 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25937768 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25937801 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25937781 | - | - |
Agilent SurePrint G3 Human CGH Microarray 4x180K | 28309 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 526365 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3396776 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_1104 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_1105 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 17586 | - | - |
Agilent Human Genome CGH + SNP Microarray | A_18_P16218702 | - | - |
Agilent Human Genome CGH + SNP Microarray | A_18_P16216566 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3396776 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_33_P3396776 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25937804 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25937812 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25937821 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25937819 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25937823 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25937787 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25937796 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25937795 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05680966 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05680976 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05680941 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05680940 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25937782 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25937789 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25937775 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25937813 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05680978 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25937811 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05680935 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25937818 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25937788 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25937826 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25937793 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05680964 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05680937 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25937802 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25937790 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25937808 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25937776 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25937799 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25937786 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25937825 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25937778 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25937792 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25937801 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25937822 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05680969 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05680955 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25937792 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25937822 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05680935 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25937787 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25937788 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25937782 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25937776 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25937811 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05680937 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25937819 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25937778 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25937825 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25937801 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05680941 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25937823 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25937796 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25937818 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25937813 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25937795 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25937808 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25937804 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25937786 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05680964 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25937799 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25937812 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25937802 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05680978 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05680976 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25937790 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25937775 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25937826 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05680940 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25937821 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05680966 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25937789 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05680955 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25937793 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05680969 | - | - |
Agilent Human Genome CGH Microarray 105A (G4412A) | A_16_P01776484 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P01776484 | - | - |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 57068 | - | - |
Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 29413 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P01776484 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05680973 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05680986 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05680959 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05680942 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05680981 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05680965 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05680953 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05680985 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05680924 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05680949 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05680927 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05680961 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16004079 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05680990 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05680989 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05680943 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05680956 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05680962 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05680936 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05680940 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05680978 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05680929 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05680988 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05680945 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05680957 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05680971 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05680938 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05680976 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05680951 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05680932 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P01776484 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | NM_001024607_at | - | - |
Affymetrix Human Gene 1.0 ST Array | 8142330 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8142331 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8142329 | - | - |
Affymetrix Human Genome U219 Array | 11732943_PM_at | - | - |
Affymetrix Human Genome U219 Array | 11732943_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | PSR07027445.hg.1 | TC07001753.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR07027445.hg.1 | ENST00000379007.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07027445.hg.1 | NM_001024607.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012242.hg.1 | ENST00000624695.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012242.hg.1 | ENST00000624695.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012242.hg.1 | NM_001024607.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012242.hg.1 | NM_001024607.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR07027444.hg.1 | TC07001753.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR07027444.hg.1 | ENST00000379007.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07027444.hg.1 | NM_001024607.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700188405.hg.1 | ENST00000624695.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700188405.hg.1 | NM_001024607.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700188405.hg.1 | ENST00000624695.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700188405.hg.1 | NM_001024607.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700188405.hg.1 | TC0700012242.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0700188406.hg.1 | ENST00000624695.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700188406.hg.1 | NM_001024607.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700188406.hg.1 | ENST00000624695.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700188406.hg.1 | NM_001024607.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700188406.hg.1 | TC0700012242.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0700012242.hg.1 | ENST00000624695.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012242.hg.1 | ENST00000624695.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012242.hg.1 | NM_001024607.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700012242.hg.1 | NM_001024607.2 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A033372 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A033372 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_1726640 | NM_001024607.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1726640 | NM_001024607.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1726640 | NM_001024607.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1726640 | NM_001024607.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1726640 | NM_001024607.1 | human_transcriptome |
Illumina HumanRef-8 v2 Expression BeadChip | GI_21029-S | - | - |
Illumina human-6 v2.0 expression beadchip | ILMN_1726640 | NM_001024607.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1726640 | NM_001024607.1 | human_transcriptome |
Illumina Sentrix Human-6 v2 Expression BeadChip | 6220703 | - | - |
Illumina HumanRef-6 v2.0 expression beadchip | ILMN_21029 | - | - |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1726640 | NM_001024607.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1726640 | NM_001024607.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1726640 | NM_001024607.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1726640 | NM_001024607.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1726640 | NM_001024607.1 | human_transcriptome |
