Assay Details
Gene Symbol: FAM47C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013736.2 | NP_001013758.1 | 1 | 1181 | 121 | |
GenBank mRNA | AK125992.1 | - | 1 | 903 | 121 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC0X000166.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11700671 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32405698 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P17333373 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32405681 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32405769 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11700657 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32405741 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11700667 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P17333437 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P17333446 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11700655 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P17333441 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32405694 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11700674 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P17333462 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11700656 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32405738 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P17333445 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11700654 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32405677 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11700659 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32405678 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32405680 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32405733 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32405683 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32405728 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11700672 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11700658 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11700670 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11700661 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11700660 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P17333442 | - | - |
Agilent SurePrint G3 Human CGH Microarray 4x180K | 83805 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 644476 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 330259 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 352563 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3249160 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_32_P158417 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 28068 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 22307 | - | - |
Agilent Human Genome CGH + SNP Microarray | A_18_P17142183 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3249160 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_33_P3249160 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32405684 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32405701 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32405698 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11700657 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32405673 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11700654 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11700656 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11700655 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11700658 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32405683 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32405679 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17333372 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32405674 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32405677 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32405669 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32405678 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32405681 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32405694 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32405702 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32405680 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32405685 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17333372 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32405680 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11700657 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32405702 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32405674 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32405685 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32405678 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32405679 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11700656 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11700658 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32405698 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11700655 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32405669 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32405701 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32405677 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32405684 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32405681 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32405683 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32405694 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32405673 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11700654 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_32_P158417 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P11700659 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P11700660 | - | - |
Agilent Whole Human Genome Microarray 4x44K G4112F | A_32_P158417 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P03689564 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11700657 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11700670 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11700667 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11700672 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11700659 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11700653 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11700651 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11700649 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11700666 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11700656 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11700674 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11700654 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11700671 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11700662 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11700668 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_18_P17142183 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | NM_001013736_at | - | - |
Affymetrix Human Gene 1.0 ST Array | 8166704 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8166703 | - | - |
Affymetrix HT HG-U133+ PM Array Plate | 239732_PM_x_at | - | - |
Affymetrix Human Genome U219 Array | 11732594_at | - | - |
Affymetrix Human Genome U219 Array | 11732594_PM_at | - | - |
Affymetrix Human Genome U219 Array | 11732595_x_at | - | - |
Affymetrix Human Genome U219 Array | 11732595_PM_x_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | PSR0X001903.hg.1 | TC0X000166.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR0X001903.hg.1 | ENST00000358047.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X001903.hg.1 | NM_001013736.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006946.hg.1 | ENST00000358047.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006946.hg.1 | ENST00000358047.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006946.hg.1 | NM_001013736.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006946.hg.1 | NM_001013736.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006946.hg.1 | NM_001013736.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR0X001902.hg.1 | TC0X000166.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR0X001902.hg.1 | ENST00000358047.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X001902.hg.1 | NM_001013736.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00147308.hg.1 | ENST00000358047.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00147308.hg.1 | NM_001013736.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00147308.hg.1 | ENST00000358047.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00147308.hg.1 | NM_001013736.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00147308.hg.1 | TC0X00006946.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0X00147306.hg.1 | NM_001013736.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00147306.hg.1 | NM_001013736.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00147306.hg.1 | TC0X00006946.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0X00006946.hg.1 | ENST00000358047.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006946.hg.1 | ENST00000358047.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006946.hg.1 | NM_001013736.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006946.hg.1 | NM_001013736.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006946.hg.1 | NM_001013736.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00147309.hg.1 | ENST00000358047.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00147309.hg.1 | NM_001013736.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00147309.hg.1 | ENST00000358047.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00147309.hg.1 | NM_001013736.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00147309.hg.1 | TC0X00006946.hg.1 | Clariom D TC |
Human Genome U133 Plus 2.0 Array | 239732_PM_x_at | - | - |
Human Genome U133 Plus 2.0 Array | 239732_x_at | - | - |
Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) | NM_001013736_at | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 239732_x_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 239732_x_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 239732_x_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 239732_x_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 239732_x_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 239732_x_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 239732_x_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 239732_x_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 239732_x_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 239732_x_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 239732_x_at6 | - | - |
Human Genome U95 Set | 72247_at | - | - |
Human Genome U95 Set | 72247_PM_at | - | - |
Human Genome U133 Set | 239732_PM_x_at | - | - |
Human Genome U133 Set | 239732_x_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A040831 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A040831 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 132481 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_1758171 | AK125992.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1758171 | NM_001013736.2 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_2071473 | AK125992.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_2071473 | NM_001013736.2 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1799721 | - | - |
HumanHT-12 V4.0 expression beadchip | ILMN_2071473 | AK125992.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_2071473 | NM_001013736.2 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1758171 | AK125992.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1758171 | NM_001013736.2 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2071473 | AK125992.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2071473 | NM_001013736.2 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2071473 | AK125992.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2071473 | NM_001013736.2 | human_transcriptome |
Illumina HumanRef-8 v2 Expression BeadChip | GI_22588-S | - | - |
Illumina human-6 v2.0 expression beadchip | ILMN_1799721 | - | - |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1799721 | - | - |
Illumina Sentrix Human-6 v2 Expression BeadChip | ILMN_22588 | - | - |
Illumina HumanRef-6 v2.0 expression beadchip | ILMN_22588 | - | - |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2071473 | AK125992.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2071473 | NM_001013736.2 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2071473 | AK125992.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2071473 | NM_001013736.2 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1758171 | AK125992.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1758171 | NM_001013736.2 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1758171 | AK125992.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1758171 | NM_001013736.2 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2071473 | AK125992.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2071473 | NM_001013736.2 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1799721 | - | - |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2071473 | AK125992.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2071473 | NM_001013736.2 | human_transcriptome |
