Assay Details
| Gene Symbol: ABCC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004996.3 | NP_004987.2 | 17 | 2321 | 87 | |
| XM_011522497.1 | XP_011520799.1 | 17 | 2137 | 87 | ||
| XM_011522498.2 | XP_011520800.1 | 16 | 2056 | 87 | ||
| XM_017023237.1 | XP_016878726.1 | 17 | 2203 | 87 | ||
| XM_017023238.1 | XP_016878727.1 | 16 | 2077 | 87 | ||
| XM_017023239.1 | XP_016878728.1 | 16 | 2065 | 87 | ||
| XM_017023241.1 | XP_016878730.1 | 15 | 1939 | 87 | ||
| XM_017023243.1 | XP_016878732.1 | 17 | 2203 | 87 | ||
| GenBank mRNA | AB209120.1 | - | 15 | 1841 | 87 | |
| CR749835.1 | - | 13 | 87 | |||
| L05628.1 | - | 17 | 2342 | 87 | ||
| X78338.1 | - | 17 | 2283 | 87 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

ABCC1
Genomic Map