Assay Details
Gene Symbol: PTPRCAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_005608.2 | NP_005599.1 | 2 | 265 | 153 | |
GenBank mRNA | AK312022.1 | - | 2 | 265 | 153 | |
BC113859.1 | - | 2 | 264 | 40030438 | 153 | |
BC114467.1 | - | 2 | 264 | 40030437 | 153 | |
CR457429.1 | - | 1 | 202 | 153 | ||
X81422.1 | - | 2 | 265 | 153 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07851021 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07851013 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07851019 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07851025 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07851014 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07851022 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07851026 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07851015 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07851024 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07851016 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07851017 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07851023 | - | - |
Agilent SurePrint G3 Human CGH Microarray 4x180K | 1652 | - | - |
Agilent SurePrint G3 Human CGH Microarray 8x60K | 62087 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3295056 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_23_P98173 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 38306 | - | - |
Agilent Human Genome CGH Microarray 44B | A_14_P114012 | - | - |
Agilent Human Genome CGH Microarray 44A | A_14_P114012 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 50956 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 60033 | - | - |
Agilent Human Genome CGH + SNP Microarray | A_14_P114012 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3295056 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_33_P3295056 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07851026 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07851015 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07851022 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07851024 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07851021 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07851017 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07851019 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07851023 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07851025 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07851015 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07851024 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07851025 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07851017 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07851023 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07851019 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07851026 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07851021 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07851022 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_23_P98173 | - | - |
Agilent Human Genome CGH Microarray 105A (G4412A) | A_14_P114012 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P07851025 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P07851026 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P07851024 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P07851021 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P07851019 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P07851017 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P07851015 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P16464801 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P16464803 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P16464799 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P07851017 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P16464802 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P07851016 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P16464800 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_14_P114012 | - | - |
Agilent Human Genome CGH Microarray 4x44K | A_14_P114012 | - | - |
Agilent Whole Human Genome Microarray 4x44K G4112F | A_23_P98173 | - | - |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 19407 | - | - |
Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 66406 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_14_P114012 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07851063 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07851024 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16464825 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07850977 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07851017 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07851026 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16464817 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07851056 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07850968 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16464831 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16464800 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07851060 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07850971 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07850987 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07850972 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07850985 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07850979 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07851023 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16464818 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07851054 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07850989 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07850970 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07851015 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07851025 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07851062 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07850974 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07851021 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07851059 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07851057 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16464802 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07850976 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07850967 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07850981 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07850983 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16464821 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07851019 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07851066 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_14_P114012 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | NM_005608_at | - | - |
Affymetrix Human Gene 1.0 ST Array | 7949793 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7949794 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7949777 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7949778 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7949795 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7949779 | - | - |
Affymetrix Human Gene 1.1 ST Array | 7949792 | - | - |
Affymetrix Human Gene 1.1 ST Array | 7949776 | - | - |
Affymetrix HT HG-U133+ PM Array Plate | 204960_PM_at | - | - |
Affymetrix Human Genome U219 Array | 11718849_at | - | - |
Affymetrix Human Genome U219 Array | 11718849_PM_at | - | - |
Affymetrix Human Clariom D Assay | PSR1100204753.hg.1 | NM_005608.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100204753.hg.1 | NM_005608.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100204753.hg.1 | TC1100013193.hg.1 | Clariom D TC |
Human Genome U133 Plus 2.0 Array | 204960_at | - | - |
Human Genome U133 Plus 2.0 Array | 204960_PM_at | - | - |
Human Genome U133A 2.0 Array | 204960_at | - | - |
Human Genome U133A 2.0 Array | 204960_PM_at | - | - |
Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) | NM_005608_at | - | - |
AffymetrixPathwork Diagnostics Pathchip | pw592856 | - | - |
Human Genome U95 Set | 32070_PM_at | - | - |
Human Genome U95 Set | 32070_at | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 204960_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 204960_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 204960_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 204960_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 204960_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 204960_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 204960_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 204960_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 204960_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 204960_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 204960_at9 | - | - |
Human Genome U133 Set | 204960_at | - | - |
Human Genome U133 Set | 204960_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A045411 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A045411 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 180800 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_1672417 | CR622472.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1672417 | NM_005608.2 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1672417 | BC113859.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1672417 | BC114467.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1672417 | CR592526.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1672417 | CR622472.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1672417 | NM_005608.2 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1672417 | BC113859.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1672417 | BC114467.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1672417 | CR592526.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1672417 | CR622472.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1672417 | NM_005608.2 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1672417 | BC113859.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1672417 | BC114467.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1672417 | CR592526.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1672417 | CR622472.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1672417 | NM_005608.2 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1672417 | BC113859.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1672417 | BC114467.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1672417 | CR592526.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1672417 | CR622472.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1672417 | NM_005608.2 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1672417 | BC113859.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1672417 | BC114467.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1672417 | CR592526.1 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_5032004-S | CR622472.1 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_5032004-S | X81422.1 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_5032004-S | NM_005608.2 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_5032004-S | BC113859.1 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_5032004-S | BC114467.1 | human_transcriptome |
Illumina HumanRef-8 v2 Expression BeadChip | GI_8267-S | - | - |
Illumina human-6 v1.0 expression beadchip | 0002060278 | - | - |
Illumina humanRef-8 v1.0 expression beadchip | 0002060278 | - | - |
Illumina human-6 v2.0 expression beadchip | ILMN_1672417 | CR622472.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1672417 | NM_005608.2 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1672417 | BC113859.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1672417 | BC114467.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1672417 | CR592526.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1672417 | CR622472.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1672417 | NM_005608.2 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1672417 | BC113859.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1672417 | BC114467.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1672417 | CR592526.1 | human_transcriptome |
Illumina Sentrix Human-6 v2 Expression BeadChip | ILMN_8267 | - | - |
Illumina HumanRef-6 v2.0 expression beadchip | ILMN_8267 | - | - |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1672417 | CR622472.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1672417 | NM_005608.2 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1672417 | BC113859.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1672417 | BC114467.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1672417 | CR592526.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1672417 | CR622472.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1672417 | NM_005608.2 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1672417 | BC113859.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1672417 | BC114467.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1672417 | CR592526.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1672417 | CR622472.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1672417 | NM_005608.2 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1672417 | BC113859.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1672417 | BC114467.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1672417 | CR592526.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1672417 | CR622472.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1672417 | NM_005608.2 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1672417 | BC113859.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1672417 | BC114467.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1672417 | CR592526.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1672417 | CR622472.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1672417 | NM_005608.2 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1672417 | BC113859.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1672417 | BC114467.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1672417 | CR592526.1 | human_transcriptome |
