Assay Details
Gene Symbol: RXRG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_033824.1 | 2 | 729 | 79 | ||
GenBank mRNA | BM684530.1 | - | 1 | 79 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC01003464.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
Function(s)
Process(es)
transcription initiation from RNA polymerase II promoter
steroid hormone mediated signaling pathway
retinoic acid receptor signaling pathway
protein homotetramerization
positive regulation of transcription from RNA polymerase II promoter involved in cellular response to chemical stimulus
RNA polymerase II regulatory region sequence-specific DNA binding
steroid hormone receptor activity
9-cis retinoic acid receptor activity
protein binding
zinc ion binding
transcription initiation from RNA polymerase II promoter
steroid hormone mediated signaling pathway
retinoic acid receptor signaling pathway
protein homotetramerization
positive regulation of transcription from RNA polymerase II promoter involved in cellular response to chemical stimulus
RNA polymerase II regulatory region sequence-specific DNA binding
steroid hormone receptor activity
9-cis retinoic acid receptor activity
protein binding
zinc ion binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR01053925.hg.1 | TC01003464.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01053925.hg.1 | NR_033824.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016254.hg.1 | ENST00000359842.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016254.hg.1 | ENST00000359842.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016254.hg.1 | ENST00000359842.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016254.hg.1 | ENST00000359842.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016254.hg.1 | ENST00000359842.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016254.hg.1 | ENST00000359842.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016254.hg.1 | ENST00000359842.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016254.hg.1 | ENST00000359842.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016254.hg.1 | ENST00000359842.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016254.hg.1 | ENST00000359842.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016254.hg.1 | ENST00000359842.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016254.hg.1 | ENST00000359842.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016254.hg.1 | ENST00000359842.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016254.hg.1 | ENST00000359842.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016254.hg.1 | ENST00000359842.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016254.hg.1 | ENST00000359842.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016254.hg.1 | ENST00000465764.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016254.hg.1 | ENST00000465764.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016254.hg.1 | ENST00000465764.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016254.hg.1 | ENST00000465764.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016254.hg.1 | ENST00000470566.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016254.hg.1 | ENST00000470566.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016254.hg.1 | ENST00000470566.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016254.hg.1 | ENST00000470566.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016254.hg.1 | ENST00000470566.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016254.hg.1 | ENST00000470566.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016254.hg.1 | ENST00000470566.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016254.hg.1 | ENST00000619224.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016254.hg.1 | ENST00000619224.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016254.hg.1 | ENST00000619224.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016254.hg.1 | ENST00000619224.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016254.hg.1 | ENST00000619224.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016254.hg.1 | ENST00000619224.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016254.hg.1 | ENST00000619224.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016254.hg.1 | ENST00000619224.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016254.hg.1 | ENST00000619224.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016254.hg.1 | ENST00000619224.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016254.hg.1 | ENST00000619224.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016254.hg.1 | ENST00000619224.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016254.hg.1 | ENST00000619224.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016254.hg.1 | ENST00000619224.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016254.hg.1 | ENST00000619224.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016254.hg.1 | ENST00000619224.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016254.hg.1 | ENST00000619224.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016254.hg.1 | ENST00000619224.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016254.hg.1 | NM_001256570.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016254.hg.1 | NM_001256570.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016254.hg.1 | NM_001256570.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016254.hg.1 | NM_001256570.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016254.hg.1 | NM_001256570.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016254.hg.1 | NM_001256570.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016254.hg.1 | NM_001256570.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016254.hg.1 | NM_001256570.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016254.hg.1 | NM_001256570.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016254.hg.1 | NM_001256570.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016254.hg.1 | NM_001256570.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016254.hg.1 | NM_001256570.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016254.hg.1 | NM_001256570.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016254.hg.1 | NM_001256570.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016254.hg.1 | NM_001256570.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016254.hg.1 | NM_001256570.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016254.hg.1 | NM_001256570.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016254.hg.1 | NM_001256570.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016254.hg.1 | NM_001256571.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016254.hg.1 | NM_001256571.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016254.hg.1 | NM_001256571.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016254.hg.1 | NM_001256571.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016254.hg.1 | NM_001256571.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016254.hg.1 | NM_001256571.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016254.hg.1 | NM_001256571.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016254.hg.1 | NM_001256571.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016254.hg.1 | NM_001256571.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016254.hg.1 | NM_001256571.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016254.hg.1 | NM_001256571.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016254.hg.1 | NM_001256571.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016254.hg.1 | NM_001256571.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016254.hg.1 | NM_001256571.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016254.hg.1 | NM_006917.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016254.hg.1 | NM_006917.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016254.hg.1 | NM_006917.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016254.hg.1 | NM_006917.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016254.hg.1 | NM_006917.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016254.hg.1 | NM_006917.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016254.hg.1 | NM_006917.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016254.hg.1 | NM_006917.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016254.hg.1 | NM_006917.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016254.hg.1 | NM_006917.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016254.hg.1 | NM_006917.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016254.hg.1 | NM_006917.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016254.hg.1 | NM_006917.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016254.hg.1 | NM_006917.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016254.hg.1 | NM_006917.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016254.hg.1 | NM_006917.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016254.hg.1 | NM_006917.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016254.hg.1 | NR_033824.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016254.hg.1 | NR_033824.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016254.hg.1 | NR_033824.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016254.hg.1 | NR_033824.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016254.hg.1 | NR_033824.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016254.hg.1 | ENST00000359842.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016254.hg.1 | ENST00000359842.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016254.hg.1 | ENST00000359842.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016254.hg.1 | ENST00000359842.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016254.hg.1 | ENST00000359842.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016254.hg.1 | ENST00000359842.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016254.hg.1 | ENST00000359842.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016254.hg.1 | ENST00000359842.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016254.hg.1 | ENST00000359842.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016254.hg.1 | ENST00000359842.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016254.hg.1 | ENST00000359842.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016254.hg.1 | ENST00000359842.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016254.hg.1 | ENST00000359842.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016254.hg.1 | ENST00000359842.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016254.hg.1 | ENST00000359842.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016254.hg.1 | ENST00000359842.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016254.hg.1 | ENST00000465764.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016254.hg.1 | ENST00000465764.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016254.hg.1 | ENST00000465764.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016254.hg.1 | ENST00000465764.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016254.hg.1 | ENST00000470566.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016254.hg.1 | ENST00000470566.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016254.hg.1 | ENST00000470566.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016254.hg.1 | ENST00000470566.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016254.hg.1 | ENST00000470566.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016254.hg.1 | ENST00000470566.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016254.hg.1 | ENST00000470566.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016254.hg.1 | ENST00000619224.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016254.hg.1 | ENST00000619224.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016254.hg.1 | ENST00000619224.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016254.hg.1 | ENST00000619224.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016254.hg.1 | ENST00000619224.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016254.hg.1 | ENST00000619224.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016254.hg.1 | ENST00000619224.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016254.hg.1 | ENST00000619224.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016254.hg.1 | ENST00000619224.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016254.hg.1 | ENST00000619224.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016254.hg.1 | ENST00000619224.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016254.hg.1 | ENST00000619224.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016254.hg.1 | ENST00000619224.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016254.hg.1 | ENST00000619224.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016254.hg.1 | ENST00000619224.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016254.hg.1 | ENST00000619224.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016254.hg.1 | ENST00000619224.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016254.hg.1 | ENST00000619224.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016254.hg.1 | NM_001256570.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016254.hg.1 | NM_001256570.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016254.hg.1 | NM_001256570.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016254.hg.1 | NM_001256570.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016254.hg.1 | NM_001256570.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016254.hg.1 | NM_001256570.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016254.hg.1 | NM_001256570.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016254.hg.1 | NM_001256570.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016254.hg.1 | NM_001256570.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016254.hg.1 | NM_001256570.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016254.hg.1 | NM_001256570.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016254.hg.1 | NM_001256570.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016254.hg.1 | NM_001256570.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016254.hg.1 | NM_001256570.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016254.hg.1 | NM_001256570.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016254.hg.1 | NM_001256570.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016254.hg.1 | NM_001256570.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016254.hg.1 | NM_001256570.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016254.hg.1 | NM_001256571.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016254.hg.1 | NM_001256571.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016254.hg.1 | NM_001256571.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016254.hg.1 | NM_001256571.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016254.hg.1 | NM_001256571.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016254.hg.1 | NM_001256571.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016254.hg.1 | NM_001256571.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016254.hg.1 | NM_001256571.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016254.hg.1 | NM_001256571.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016254.hg.1 | NM_001256571.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016254.hg.1 | NM_001256571.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016254.hg.1 | NM_001256571.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016254.hg.1 | NM_001256571.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016254.hg.1 | NM_001256571.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016254.hg.1 | NM_006917.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016254.hg.1 | NM_006917.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016254.hg.1 | NM_006917.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016254.hg.1 | NM_006917.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016254.hg.1 | NM_006917.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016254.hg.1 | NM_006917.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016254.hg.1 | NM_006917.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016254.hg.1 | NM_006917.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016254.hg.1 | NM_006917.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016254.hg.1 | NM_006917.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016254.hg.1 | NM_006917.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016254.hg.1 | NM_006917.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016254.hg.1 | NM_006917.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016254.hg.1 | NM_006917.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016254.hg.1 | NM_006917.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016254.hg.1 | NM_006917.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016254.hg.1 | NM_006917.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016254.hg.1 | NR_033824.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016254.hg.1 | NR_033824.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016254.hg.1 | NR_033824.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016254.hg.1 | NR_033824.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016254.hg.1 | NR_033824.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100223708.hg.1 | ENST00000465764.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100223708.hg.1 | NR_033824.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100223708.hg.1 | ENST00000465764.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100223708.hg.1 | NR_033824.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100223708.hg.1 | TC0100016254.hg.1 | Clariom D TC |
