Assay Details
| Gene Symbol: DDT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084392.1 | NP_001077861.1 | 3 | 504 | 69 | |
| NM_001355.3 | NP_001346.1 | 4 | 555 | 69 | ||
| GenBank mRNA | AK292044.1 | - | 3 | 448 | 69 | |
| BC005971.1 | - | 3 | 452 | 4082777 | 69 | |
| BC015508.2 | - | 3 | 473 | 3902943 | 69 | |
| CA414359.1 | - | 1 | 69 | |||
| U49785.1 | - | 4 | 553 | 69 | ||
| U84143.1 | - | 3 | 441 | 69 | ||
| Gene Symbol: COX8A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC000501.1 | - | 605 | 69 | ||
| Gene Symbol: DDTL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084393.1 | NP_001077862.1 | 3 | 69 | ||
| GenBank mRNA | AK296110.1 | - | 3 | 69 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC22000141.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32122679 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32122676 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32122678 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32122674 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11444680 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P17769 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_23_P17769 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 8206 | - | - |
| Agilent Human Genome CGH Microarray 44B | A_14_P138267 | - | - |
| Agilent Human Genome CGH Microarray 44B | A_14_P112167 | - | - |
| Agilent Human Genome CGH Microarray 44A | A_14_P138267 | - | - |
| Agilent Human Genome CGH Microarray 44A | A_14_P112167 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 26538 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P17769 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_23_P17769 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11444680 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32122678 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32122679 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32122676 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32122674 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32122674 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32122678 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11444680 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32122676 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32122679 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_23_P17769 | - | - |
| Agilent Human Genome CGH Microarray 105A (G4412A) | A_14_P138267 | - | - |
| Agilent Human Genome CGH Microarray 105A (G4412A) | A_14_P112167 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_14_P138267 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P21291596 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_14_P112167 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_23_P17769 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11444677 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11444670 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11444683 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11444680 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11444666 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11444673 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11444668 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11444667 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | NM_001084392_at | - | - |
| Affymetrix Human Gene 1.0 ST Array | NM_001355_at | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8071808 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8074970 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8074971 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8071807 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8071806 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8074969 | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 202929_PM_s_at | - | - |
| Affymetrix Human Genome U219 Array | 11731089_a_at | - | - |
| Affymetrix Human Genome U219 Array | 11731089_PM_a_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | PSR22002578.hg.1 | NM_001084393.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22002578.hg.1 | TC22000141.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006874.hg.1 | ENST00000215770.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006874.hg.1 | ENST00000215770.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006874.hg.1 | ENST00000215770.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006874.hg.1 | ENST00000215770.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006874.hg.1 | NM_001084393.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006874.hg.1 | NM_001084393.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006874.hg.1 | NM_001084393.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006874.hg.1 | NM_001084393.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006874.hg.1 | NM_001084393.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006874.hg.1 | NM_001084393.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006874.hg.1 | NM_001144931.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006874.hg.1 | NM_001144931.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006874.hg.1 | NM_001355.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006874.hg.1 | NM_001355.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006874.hg.1 | NM_001355.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200147478.hg.1 | TC2200006874.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200147478.hg.1 | NM_001084393.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200147478.hg.1 | NM_001084393.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006874.hg.1 | ENST00000215770.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006874.hg.1 | ENST00000215770.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006874.hg.1 | ENST00000215770.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006874.hg.1 | ENST00000215770.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006874.hg.1 | NM_001084393.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006874.hg.1 | NM_001084393.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006874.hg.1 | NM_001084393.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006874.hg.1 | NM_001084393.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006874.hg.1 | NM_001084393.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006874.hg.1 | NM_001084393.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006874.hg.1 | NM_001144931.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006874.hg.1 | NM_001144931.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006874.hg.1 | NM_001355.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006874.hg.1 | NM_001355.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006874.hg.1 | NM_001355.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200147477.hg.1 | TC2200006874.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200147477.hg.1 | ENST00000215770.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200147477.hg.1 | NM_001084393.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200147477.hg.1 | ENST00000215770.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200147477.hg.1 | NM_001084393.1 | Refseq |
| Human Genome U133 Plus 2.0 Array | 202929_PM_s_at | - | - |
| Human Genome U133 Plus 2.0 Array | 202929_s_at | - | - |
| Human Genome U133A 2.0 Array | 202929_PM_s_at | - | - |
| Human Genome U133A 2.0 Array | 202929_s_at | - | - |
| Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) | NM_001355_at | - | - |
| AffymetrixPathwork Diagnostics Pathchip | pw571571 | - | - |
| Human Genome U95 Set | 33689_s_at | - | - |
| Human Genome U95 Set | 41181_r_at | - | - |
| Human Genome U95 Set | 41180_i_at | - | - |
| Human Genome U95 Set | 374_f_at | - | - |
| Human Genome U95 Set | 33689_PM_s_at | - | - |
| Human Genome U95 Set | 41181_PM_r_at | - | - |
| Human Genome U95 Set | 374_PM_f_at | - | - |
| Human Genome U95 Set | 41180_PM_i_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 202929_s_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 202929_s_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 202929_s_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 202929_s_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 202929_s_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 202929_s_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 202929_s_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 202929_s_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 202929_s_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 202929_s_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 202929_s_at7 | - | - |
| Human Genome U133 Set | 202929_PM_s_at | - | - |
| Human Genome U133 Set | 202929_s_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A019579 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A019577 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A019577 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A019579 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 136242 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_1690982 | AK296110.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1690982 | CR590354.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1690982 | CR606018.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1690982 | NM_001084393.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1690982 | NM_001084392.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1690982 | AK296110.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1690982 | CR590354.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1690982 | CR606018.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1690982 | NM_001084393.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1690982 | NM_001084392.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1690982 | AK296110.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1690982 | CR590354.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1690982 | CR606018.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1690982 | NM_001084393.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1690982 | NM_001084392.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1690982 | AK296110.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1690982 | CR590354.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1690982 | CR606018.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1690982 | NM_001084393.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1690982 | NM_001084392.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1690982 | AK296110.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1690982 | CR590354.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1690982 | CR606018.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1690982 | NM_001084393.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1690982 | NM_001084392.1 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_5453630-S | AK296110.1 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_5453630-S | CR590354.1 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_5453630-S | CR606018.1 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_5453630-S | NM_001084393.1 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_5453630-S | NM_001084392.1 | human_transcriptome |
| Illumina HumanRef-8 v2 Expression BeadChip | GI_24330-S | - | - |
| Illumina human-6 v1.0 expression beadchip | 0006510632 | - | - |
| Illumina humanRef-8 v1.0 expression beadchip | 0006510632 | - | - |
| Illumina human-6 v2.0 expression beadchip | ILMN_1690982 | AK296110.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1690982 | CR590354.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1690982 | CR606018.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1690982 | NM_001084393.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1690982 | NM_001084392.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1690982 | AK296110.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1690982 | CR590354.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1690982 | CR606018.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1690982 | NM_001084393.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1690982 | NM_001084392.1 | human_transcriptome |
| Illumina Sentrix Human-6 v2 Expression BeadChip | ILMN_24330 | - | - |
| Illumina HumanRef-6 v2.0 expression beadchip | ILMN_24330 | - | - |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1690982 | AK296110.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1690982 | CR590354.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1690982 | CR606018.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1690982 | NM_001084393.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1690982 | NM_001084392.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1690982 | AK296110.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1690982 | CR590354.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1690982 | CR606018.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1690982 | NM_001084393.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1690982 | NM_001084392.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1690982 | AK296110.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1690982 | CR590354.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1690982 | CR606018.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1690982 | NM_001084393.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1690982 | NM_001084392.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1690982 | AK296110.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1690982 | CR590354.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1690982 | CR606018.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1690982 | NM_001084393.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1690982 | NM_001084392.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1690982 | AK296110.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1690982 | CR590354.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1690982 | CR606018.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1690982 | NM_001084393.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1690982 | NM_001084392.1 | human_transcriptome |

DDT
Genomic Map