Assay Details
Gene Symbol: SREBF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005291.2 | NP_001005291.1 | 12 | 2448 | 55 | |
NM_001321096.2 | NP_001308025.1 | 11 | 2220 | 55 | ||
NM_004176.4 | NP_004167.3 | 11 | 2358 | 55 | ||
XM_005256772.4 | XP_005256829.1 | 11 | 2358 | 55 | ||
XM_017024970.1 | XP_016880459.1 | 9 | 1789 | 55 | ||
XM_017024971.1 | XP_016880460.1 | 7 | 1140 | 55 | ||
GenBank mRNA | AB209609.1 | - | 10 | 2824 | 55 | |
AK091131.1 | - | 11 | 2187 | 55 | ||
AK095325.1 | - | 6 | 1197 | 55 | ||
AK128320.1 | - | 9 | 1867 | 55 | ||
AK293795.1 | - | 11 | 2058 | 55 | ||
AK294800.1 | - | 12 | 2036 | 55 | ||
AK297113.1 | - | 9 | 1772 | 55 | ||
BC023621.2 | - | 5 | 782 | 55 | ||
BC026962.1 | - | 5 | 974 | 55 | ||
BC057388.1 | - | 11 | 2349 | 5786483 | 55 | |
BC063281.1 | - | 12 | 2423 | 5264244 | 55 | |
U00968.1 | - | 11 | 2330 | 55 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
