| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14000161.hg.1 |
ENST00000315915.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14000161.hg.1 |
NM_001005483.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14000161.hg.1 |
TC14000038.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400006491.hg.1 |
ENST00000315915.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400006491.hg.1 |
ENST00000315915.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400006491.hg.1 |
ENST00000315915.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400006491.hg.1 |
NM_001005483.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400144316.hg.1 |
ENST00000315915.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400144316.hg.1 |
NM_001005483.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400144316.hg.1 |
ENST00000315915.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400144316.hg.1 |
NM_001005483.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400144316.hg.1 |
TC1400006491.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1400006491.hg.1 |
ENST00000315915.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400006491.hg.1 |
ENST00000315915.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400006491.hg.1 |
ENST00000315915.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400006491.hg.1 |
NM_001005483.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400144317.hg.1 |
ENST00000315915.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400144317.hg.1 |
ENST00000315915.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400144317.hg.1 |
TC1400006491.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1400144315.hg.1 |
ENST00000315915.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400144315.hg.1 |
ENST00000315915.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400144315.hg.1 |
TC1400006491.hg.1 |
Clariom D TC |