Assay Details
Gene Symbol: CLSTN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022131.2 | NP_071414.2 | 17 | 4547 | 64 | |
XM_017007022.1 | XP_016862511.1 | 17 | 4588 | 64 | ||
GenBank mRNA | AJ278018.1 | - | 18 | 4367 | 64 | |
AK096818.1 | - | 2 | 1882 | 64 | ||
BC004871.2 | - | 5 | 2209 | 64 | ||
BC007943.2 | - | 3 | 1998 | 64 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000511524.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000511524.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000511524.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000511524.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000511524.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000511524.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000511524.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000511524.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000511524.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000511524.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000511524.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000511524.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000511524.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000458420.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000511524.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000511524.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000511524.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000511524.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000511524.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000511524.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000511524.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000511524.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000511524.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000511524.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000511524.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000511524.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000511524.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | ENST00000620185.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008979.hg.1 | NM_022131.2 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 191167 | - | - |
