Assay Details
| Gene Symbol: ASCC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006828.3 | NP_006819.2 | 17 - 18 | 3170 | 132 | |
| XM_011535394.2 | XP_011533696.1 | 17 - 18 | 3180 | 132 | ||
| XM_011535395.2 | XP_011533697.1 | 18 - 19 | 3405 | 132 | ||
| XM_011535396.2 | XP_011533698.1 | 16 - 17 | 3014 | 132 | ||
| XM_017010205.1 | XP_016865694.1 | 18 - 19 | 3426 | 132 | ||
| XM_017010206.1 | XP_016865695.1 | 18 - 19 | 3204 | 132 | ||
| GenBank mRNA | AK299117.1 | - | 16 - 17 | 3019 | 132 | |
| AL834463.1 | - | 16 - 17 | 2855 | 132 | ||
| AY013288.1 | - | 13 - 14 | 1989 | 132 | ||
More Information
Set Membership: |
|

ASCC3
Genomic Map