Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10023205.hg.1 |
ENST00000522781.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10023205.hg.1 |
NM_001039762.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10023205.hg.1 |
TC10001752.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR10023209.hg.1 |
ENST00000522781.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10023209.hg.1 |
NM_001039762.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10023209.hg.1 |
TC10001752.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012213.hg.1 |
ENST00000424811.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012213.hg.1 |
ENST00000424811.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012213.hg.1 |
ENST00000424811.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012213.hg.1 |
ENST00000424811.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012213.hg.1 |
ENST00000424811.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012213.hg.1 |
ENST00000522781.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012213.hg.1 |
ENST00000522781.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012213.hg.1 |
ENST00000522781.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012213.hg.1 |
ENST00000522781.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012213.hg.1 |
ENST00000522781.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012213.hg.1 |
ENST00000522781.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012213.hg.1 |
ENST00000522781.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012213.hg.1 |
ENST00000522781.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012213.hg.1 |
ENST00000522781.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012213.hg.1 |
ENST00000522781.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012213.hg.1 |
ENST00000614311.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012213.hg.1 |
ENST00000614311.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012213.hg.1 |
ENST00000614311.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012213.hg.1 |
ENST00000614311.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012213.hg.1 |
ENST00000614311.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012213.hg.1 |
ENST00000614311.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012213.hg.1 |
ENST00000614311.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012213.hg.1 |
ENST00000614311.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012213.hg.1 |
ENST00000614311.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012213.hg.1 |
NM_001039762.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012213.hg.1 |
NM_001039762.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012213.hg.1 |
NM_001039762.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012213.hg.1 |
NM_001039762.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012213.hg.1 |
NM_001039762.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012213.hg.1 |
NM_001039762.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012213.hg.1 |
NM_001039762.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012213.hg.1 |
NM_001039762.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012213.hg.1 |
NM_001039762.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012213.hg.1 |
NM_001039762.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10023208.hg.1 |
ENST00000424811.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10023208.hg.1 |
ENST00000522781.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10023208.hg.1 |
NM_001039762.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10023208.hg.1 |
TC10001752.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR10023210.hg.1 |
ENST00000522781.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10023210.hg.1 |
NM_001039762.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10023210.hg.1 |
TC10001752.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR10023206.hg.1 |
ENST00000424811.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10023206.hg.1 |
ENST00000522781.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10023206.hg.1 |
NM_001039762.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10023206.hg.1 |
TC10001752.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR10023211.hg.1 |
ENST00000522781.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10023211.hg.1 |
NM_001039762.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10023211.hg.1 |
TC10001752.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1000183478.hg.1 |
ENST00000522781.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000183478.hg.1 |
ENST00000614311.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000183478.hg.1 |
NM_001039762.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000183478.hg.1 |
ENST00000522781.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000183478.hg.1 |
ENST00000614311.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000183478.hg.1 |
NM_001039762.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000183478.hg.1 |
TC1000012213.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000183477.hg.1 |
ENST00000522781.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000183477.hg.1 |
ENST00000614311.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000183477.hg.1 |
NM_001039762.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000183477.hg.1 |
ENST00000522781.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000183477.hg.1 |
ENST00000614311.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000183477.hg.1 |
NM_001039762.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000183477.hg.1 |
TC1000012213.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000183476.hg.1 |
ENST00000424811.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000183476.hg.1 |
ENST00000522781.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000183476.hg.1 |
ENST00000614311.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000183476.hg.1 |
NM_001039762.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000183476.hg.1 |
ENST00000424811.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000183476.hg.1 |
ENST00000522781.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000183476.hg.1 |
ENST00000614311.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000183476.hg.1 |
NM_001039762.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000183476.hg.1 |
TC1000012213.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1000012213.hg.1 |
ENST00000424811.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012213.hg.1 |
ENST00000424811.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012213.hg.1 |
ENST00000424811.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012213.hg.1 |
ENST00000424811.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012213.hg.1 |
ENST00000424811.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012213.hg.1 |
ENST00000522781.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012213.hg.1 |
ENST00000522781.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012213.hg.1 |
ENST00000522781.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012213.hg.1 |
ENST00000522781.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012213.hg.1 |
ENST00000522781.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012213.hg.1 |
ENST00000522781.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012213.hg.1 |
ENST00000522781.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012213.hg.1 |
ENST00000522781.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012213.hg.1 |
ENST00000522781.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012213.hg.1 |
ENST00000522781.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012213.hg.1 |
ENST00000614311.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012213.hg.1 |
ENST00000614311.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012213.hg.1 |
ENST00000614311.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012213.hg.1 |
ENST00000614311.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012213.hg.1 |
ENST00000614311.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012213.hg.1 |
ENST00000614311.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012213.hg.1 |
ENST00000614311.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012213.hg.1 |
ENST00000614311.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012213.hg.1 |
ENST00000614311.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012213.hg.1 |
NM_001039762.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012213.hg.1 |
NM_001039762.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012213.hg.1 |
NM_001039762.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012213.hg.1 |
NM_001039762.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012213.hg.1 |
NM_001039762.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012213.hg.1 |
NM_001039762.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012213.hg.1 |
NM_001039762.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012213.hg.1 |
NM_001039762.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012213.hg.1 |
NM_001039762.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012213.hg.1 |
NM_001039762.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000183470.hg.1 |
ENST00000522781.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000183470.hg.1 |
NM_001039762.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000183470.hg.1 |
ENST00000522781.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000183470.hg.1 |
NM_001039762.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000183470.hg.1 |
TC1000012213.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000183472.hg.1 |
ENST00000424811.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000183472.hg.1 |
ENST00000522781.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000183472.hg.1 |
ENST00000614311.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000183472.hg.1 |
NM_001039762.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000183472.hg.1 |
ENST00000424811.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000183472.hg.1 |
ENST00000522781.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000183472.hg.1 |
ENST00000614311.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000183472.hg.1 |
NM_001039762.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000183472.hg.1 |
TC1000012213.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000183479.hg.1 |
ENST00000522781.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000183479.hg.1 |
ENST00000614311.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000183479.hg.1 |
NM_001039762.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000183479.hg.1 |
ENST00000522781.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000183479.hg.1 |
ENST00000614311.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000183479.hg.1 |
NM_001039762.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000183479.hg.1 |
TC1000012213.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000183474.hg.1 |
TC1000012213.hg.1 |
Clariom D TC |