Assay Details
Gene Symbol: HTATSF1P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_033884.1 | 1 | 1507 | 82 | ||
GenBank mRNA | AK128391.1 | - | 1 | 632 | 82 | |
BX648511.1 | - | 1 | 627 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 434774 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 342 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 84748 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 39229 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 20608 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 6802 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_32_P135890 | - | - |
Agilent Whole Human Genome Microarray 4x44K G4112F | A_32_P135890 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P24734131 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P15522406 | - | - |
Agilent Human 1B Microarray G4111A | A_32_P163156 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_18_P15522406 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | 8123638 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8123640 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8123639 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8116619 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8123641 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8123637 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8116610 | - | - |
Affymetrix HT HG-U133+ PM Array Plate | 1558882_PM_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | TC0600014222.hg.1 | ENST00000604204.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014222.hg.1 | ENST00000604204.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014222.hg.1 | NR_033884.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014222.hg.1 | NR_033884.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014222.hg.1 | NR_033884.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014222.hg.1 | NR_033884.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014222.hg.1 | NR_033884.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014222.hg.1 | NR_033884.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014222.hg.1 | NR_033884.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014222.hg.1 | NR_033884.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014222.hg.1 | NR_033884.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014222.hg.1 | NR_033884.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014222.hg.1 | NR_033884.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014056.hg.1 | ENST00000603222.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600200320.hg.1 | NR_033884.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600200320.hg.1 | NR_033884.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600200320.hg.1 | TC0600014222.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0600200321.hg.1 | NR_033884.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600200321.hg.1 | NR_033884.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600200321.hg.1 | TC0600014222.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0600200326.hg.1 | NR_033884.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600200326.hg.1 | NR_033884.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600200326.hg.1 | TC0600014222.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0600014056.hg.1 | ENST00000603222.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600200319.hg.1 | NR_033884.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600200319.hg.1 | NR_033884.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600200319.hg.1 | TC0600014222.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0600200322.hg.1 | NR_033884.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600200322.hg.1 | NR_033884.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600200322.hg.1 | TC0600014222.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0600200325.hg.1 | ENST00000604204.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600200325.hg.1 | NR_033884.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600200325.hg.1 | ENST00000604204.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600200325.hg.1 | NR_033884.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600200325.hg.1 | TC0600014222.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0600200328.hg.1 | NR_033884.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600200328.hg.1 | NR_033884.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600200328.hg.1 | TC0600014222.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0600014222.hg.1 | ENST00000604204.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014222.hg.1 | ENST00000604204.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014222.hg.1 | NR_033884.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014222.hg.1 | NR_033884.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014222.hg.1 | NR_033884.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014222.hg.1 | NR_033884.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014222.hg.1 | NR_033884.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014222.hg.1 | NR_033884.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014222.hg.1 | NR_033884.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014222.hg.1 | NR_033884.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014222.hg.1 | NR_033884.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014222.hg.1 | NR_033884.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014222.hg.1 | NR_033884.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600200318.hg.1 | NR_033884.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600200318.hg.1 | NR_033884.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600200318.hg.1 | TC0600014222.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0600200323.hg.1 | NR_033884.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600200323.hg.1 | NR_033884.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600200323.hg.1 | TC0600014222.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0600200327.hg.1 | NR_033884.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600200327.hg.1 | NR_033884.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600200327.hg.1 | TC0600014222.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0600200324.hg.1 | ENST00000604204.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600200324.hg.1 | NR_033884.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600200324.hg.1 | ENST00000604204.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600200324.hg.1 | NR_033884.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600200324.hg.1 | TC0600014222.hg.1 | Clariom D TC |
Human Genome U133 Plus 2.0 Array | 1558882_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 1558882_at | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1558882_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1558882_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1558882_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1558882_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1558882_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1558882_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1558882_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1558882_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1558882_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1558882_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1558882_at10 | - | - |
Human Genome U95 Set | 49657_at | - | - |
Human Genome U95 Set | 49657_PM_at | - | - |
Human Genome U95 Set | 56963_PM_f_at | - | - |
Human Genome U95 Set | 56963_f_at | - | - |
Human Genome U95 Set | 73248_PM_at | - | - |
Human Genome U95 Set | 73248_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A029329 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A029329 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_1674285 | AK128391.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_3307276 | - | - |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1674285 | AK128391.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1674285 | AK128391.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1674285 | AK128391.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1674285 | AK128391.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1674285 | AK128391.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1674285 | AK128391.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1674285 | AK128391.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1674285 | AK128391.1 | human_transcriptome |
