Assay Details
Gene Symbol: USP27X-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_026742.1 | 2 | 385 | 144 | ||
GenBank mRNA | AK056746.1 | - | 2 | 271 | 144 | |
BC093943.1 | - | 2 | 145 | 144 | ||
BC093945.1 | - | 2 | 146 | 144 | ||
DA687298.1 | - | 2 | 385 | 144 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC0X001033.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11744690 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11744692 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11744694 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32453072 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11744676 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32453071 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11744672 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11744675 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11744678 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11744673 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11744697 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32453068 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11744684 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11744671 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32453086 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32453066 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11744696 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32453076 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32453067 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32453069 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11744680 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11744688 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32453075 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 139325 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_32_P155826 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_4746 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_4747 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 37882 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 12540 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_32_P155826 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_32_P155826 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11744690 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11744692 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32453067 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11744691 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11744675 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11744696 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32453066 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32453072 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11744671 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11744672 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32453074 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11744678 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11744673 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32453076 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32453071 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32453070 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11744688 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32453057 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11744676 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32453068 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11744694 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32453075 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32453069 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32453069 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11744692 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11744675 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11744671 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11744678 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11744673 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11744690 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32453072 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32453068 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32453071 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11744694 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11744672 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32453070 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32453075 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11744688 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11744676 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32453067 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32453074 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32453057 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11744691 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32453066 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11744696 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32453076 | - | - |
Agilent Human 1B Microarray G4111A | A_32_P521819 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11744687 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11744686 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11744682 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11744675 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11744693 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P17342749 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11744696 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11744694 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P17342758 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11744688 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11744690 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11744684 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P17342762 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11744673 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11744681 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P17342764 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11744678 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P17342756 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P17342759 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | 8172655 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8172654 | - | - |
Affymetrix HT HG-U133+ PM Array Plate | 243017_PM_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | PSR0X013917.hg.1 | TC0X001033.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR0X013917.hg.1 | ENST00000437322.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X013917.hg.1 | NR_026742.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR0X013916.hg.1 | TC0X001033.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR0X013916.hg.1 | ENST00000437322.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X013916.hg.1 | NR_026742.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR0X013918.hg.1 | TC0X001033.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR0X013918.hg.1 | NR_026742.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009673.hg.1 | ENST00000437322.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009673.hg.1 | ENST00000437322.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009673.hg.1 | ENST00000437322.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009673.hg.1 | NR_026742.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009673.hg.1 | NR_026742.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009673.hg.1 | NR_026742.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009673.hg.1 | NR_026742.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR0X013915.hg.1 | TC0X001033.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR0X013915.hg.1 | NR_026742.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00166103.hg.1 | NR_026742.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00166103.hg.1 | NR_026742.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00166103.hg.1 | TC0X00009673.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0X00166100.hg.1 | ENST00000437322.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00166100.hg.1 | NR_026742.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00166100.hg.1 | ENST00000437322.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00166100.hg.1 | NR_026742.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00166100.hg.1 | TC0X00009673.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0X00166101.hg.1 | ENST00000437322.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00166101.hg.1 | NR_026742.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00166101.hg.1 | ENST00000437322.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00166101.hg.1 | NR_026742.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00166101.hg.1 | TC0X00009673.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0X00009673.hg.1 | ENST00000437322.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009673.hg.1 | ENST00000437322.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009673.hg.1 | ENST00000437322.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009673.hg.1 | NR_026742.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009673.hg.1 | NR_026742.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009673.hg.1 | NR_026742.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009673.hg.1 | NR_026742.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00166102.hg.1 | ENST00000437322.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00166102.hg.1 | NR_026742.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00166102.hg.1 | ENST00000437322.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00166102.hg.1 | NR_026742.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00166102.hg.1 | TC0X00009673.hg.1 | Clariom D TC |
Human Genome U133 Plus 2.0 Array | 243017_at | - | - |
Human Genome U133 Plus 2.0 Array | 243017_PM_at | - | - |
Human Genome U95 Set | 46023_PM_at | - | - |
Human Genome U95 Set | 46023_at | - | - |
Human Genome U133 Set | 243017_PM_at | - | - |
Human Genome U133 Set | 243017_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A039874 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A039875 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A039875 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A039874 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 228290 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_1762545 | NR_026742.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1762545 | AK056746.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1762545 | BC093943.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1762545 | BC093945.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1762545 | NR_026742.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1762545 | AK056746.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1762545 | BC093943.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1762545 | BC093945.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1762545 | NR_026742.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1762545 | AK056746.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1762545 | BC093943.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1762545 | BC093945.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1762545 | NR_026742.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1762545 | AK056746.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1762545 | BC093943.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1762545 | BC093945.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1762545 | NR_026742.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1762545 | AK056746.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1762545 | BC093943.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1762545 | BC093945.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1762545 | NR_026742.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1762545 | AK056746.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1762545 | BC093943.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1762545 | BC093945.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1762545 | NR_026742.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1762545 | AK056746.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1762545 | BC093943.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1762545 | BC093945.1 | human_transcriptome |
