Assay Details
| Gene Symbol: SPI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080547.1 | NP_001074016.1 | 3 - 4 | 553 | 85 | |
| NM_003120.2 | NP_003111.2 | 3 - 4 | 550 | 85 | ||
| XM_011520307.1 | XP_011518609.1 | 5 - 6 | 1995 | 85 | ||
| XM_017018173.1 | XP_016873662.1 | 5 - 6 | 1998 | 85 | ||
| GenBank mRNA | AK310065.1 | - | 3 - 4 | 461 | 85 | |
| BI909430.1 | - | 3 - 4 | 354 | 85 | ||
| X52056.1 | - | 3 - 4 | 520 | 85 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11001736.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
lymphoid progenitor cell differentiation
transcription from RNA polymerase II promoter
lymphocyte differentiation
erythrocyte differentiation
macrophage differentiation
granulocyte differentiation
somatic stem cell population maintenance
myeloid dendritic cell differentiation
histone H3 acetylation
hypermethylation of CpG island
negative regulation of MHC class II biosynthetic process
regulation of erythrocyte differentiation
negative regulation of gene expression, epigenetic
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
anatomical structure regression
cellular response to ethanol
negative regulation of histone H4 acetylation
apoptotic process involved in patterning of blood vessels
positive regulation of pri-miRNA transcription from RNA polymerase II promoter
RNA polymerase II core promoter proximal region sequence-specific DNA binding
RNA polymerase II core promoter sequence-specific DNA binding
RNA polymerase II distal enhancer sequence-specific DNA binding
RNA polymerase II transcription factor activity, sequence-specific DNA binding
core promoter binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
RNA polymerase II transcription factor binding
transcriptional activator activity, RNA polymerase II distal enhancer sequence-specific binding
transcription factor activity, sequence-specific DNA binding
transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding
RNA binding
protein binding
NFAT protein binding
negative regulation of transcription from RNA polymerase II promoter
lymphoid progenitor cell differentiation
transcription from RNA polymerase II promoter
lymphocyte differentiation
erythrocyte differentiation
macrophage differentiation
granulocyte differentiation
somatic stem cell population maintenance
myeloid dendritic cell differentiation
histone H3 acetylation
hypermethylation of CpG island
negative regulation of MHC class II biosynthetic process
regulation of erythrocyte differentiation
negative regulation of gene expression, epigenetic
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
anatomical structure regression
cellular response to ethanol
negative regulation of histone H4 acetylation
apoptotic process involved in patterning of blood vessels
positive regulation of pri-miRNA transcription from RNA polymerase II promoter
RNA polymerase II core promoter proximal region sequence-specific DNA binding
RNA polymerase II core promoter sequence-specific DNA binding
RNA polymerase II distal enhancer sequence-specific DNA binding
RNA polymerase II transcription factor activity, sequence-specific DNA binding
core promoter binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
RNA polymerase II transcription factor binding
transcriptional activator activity, RNA polymerase II distal enhancer sequence-specific binding
transcription factor activity, sequence-specific DNA binding
transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding
RNA binding
protein binding
NFAT protein binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1100010733.hg.1 | ENST00000227163.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010733.hg.1 | ENST00000227163.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010733.hg.1 | ENST00000227163.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010733.hg.1 | ENST00000227163.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010733.hg.1 | ENST00000227163.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010733.hg.1 | ENST00000227163.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010733.hg.1 | ENST00000227163.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010733.hg.1 | ENST00000227163.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010733.hg.1 | ENST00000378538.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010733.hg.1 | ENST00000378538.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010733.hg.1 | ENST00000378538.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010733.hg.1 | ENST00000378538.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010733.hg.1 | ENST00000378538.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010733.hg.1 | ENST00000378538.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010733.hg.1 | ENST00000378538.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010733.hg.1 | ENST00000378538.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010733.hg.1 | ENST00000378538.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010733.hg.1 | ENST00000378538.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010733.hg.1 | ENST00000378538.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010733.hg.1 | ENST00000378538.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010733.hg.1 | ENST00000533030.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010733.hg.1 | ENST00000533030.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010733.hg.1 | ENST00000533030.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010733.hg.1 | ENST00000533030.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010733.hg.1 | ENST00000533030.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010733.hg.1 | ENST00000533030.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010733.hg.1 | ENST00000533968.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010733.hg.1 | ENST00000533968.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010733.hg.1 | ENST00000533968.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010733.hg.1 | ENST00000533968.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010733.hg.1 | ENST00000533968.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010733.hg.1 | ENST00000533968.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010733.hg.1 | ENST00000533968.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010733.hg.1 | ENST00000533968.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010733.hg.1 | NM_001080547.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010733.hg.1 | NM_001080547.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010733.hg.1 | NM_001080547.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010733.hg.1 | NM_001080547.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010733.hg.1 | NM_001080547.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010733.hg.1 | NM_001080547.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010733.hg.1 | NM_001080547.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010733.hg.1 | NM_001080547.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010733.hg.1 | NM_001080547.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010733.hg.1 | NM_001080547.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010733.hg.1 | NM_001080547.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010733.hg.1 | NM_001080547.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010733.hg.1 | NM_003120.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010733.hg.1 | NM_003120.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010733.hg.1 | NM_003120.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010733.hg.1 | NM_003120.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010733.hg.1 | NM_003120.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010733.hg.1 | NM_003120.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010733.hg.1 | NM_003120.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010733.hg.1 | NM_003120.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010733.hg.1 | NM_003120.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010733.hg.1 | NM_003120.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010733.hg.1 | NM_003120.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010733.hg.1 | NM_003120.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11020299.hg.1 | ENST00000227163.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11020299.hg.1 | ENST00000378538.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11020299.hg.1 | ENST00000533968.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11020299.hg.1 | NM_001080547.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11020299.hg.1 | NM_003120.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11020299.hg.1 | TC11001736.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1100181357.hg.1 | ENST00000227163.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100181357.hg.1 | ENST00000378538.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100181357.hg.1 | ENST00000533968.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100181357.hg.1 | NM_001080547.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100181357.hg.1 | NM_003120.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100181357.hg.1 | ENST00000227163.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100181357.hg.1 | ENST00000378538.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100181357.hg.1 | ENST00000533968.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100181357.hg.1 | NM_001080547.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100181357.hg.1 | NM_003120.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100181357.hg.1 | TC1100010733.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1100010733.hg.1 | ENST00000227163.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010733.hg.1 | ENST00000227163.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010733.hg.1 | ENST00000227163.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010733.hg.1 | ENST00000227163.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010733.hg.1 | ENST00000227163.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010733.hg.1 | ENST00000227163.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010733.hg.1 | ENST00000227163.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010733.hg.1 | ENST00000227163.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010733.hg.1 | ENST00000378538.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010733.hg.1 | ENST00000378538.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010733.hg.1 | ENST00000378538.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010733.hg.1 | ENST00000378538.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010733.hg.1 | ENST00000378538.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010733.hg.1 | ENST00000378538.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010733.hg.1 | ENST00000378538.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010733.hg.1 | ENST00000378538.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010733.hg.1 | ENST00000378538.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010733.hg.1 | ENST00000378538.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010733.hg.1 | ENST00000378538.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010733.hg.1 | ENST00000378538.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010733.hg.1 | ENST00000533030.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010733.hg.1 | ENST00000533030.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010733.hg.1 | ENST00000533030.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010733.hg.1 | ENST00000533030.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010733.hg.1 | ENST00000533030.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010733.hg.1 | ENST00000533030.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010733.hg.1 | ENST00000533968.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010733.hg.1 | ENST00000533968.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010733.hg.1 | ENST00000533968.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010733.hg.1 | ENST00000533968.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010733.hg.1 | ENST00000533968.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010733.hg.1 | ENST00000533968.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010733.hg.1 | ENST00000533968.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010733.hg.1 | ENST00000533968.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010733.hg.1 | NM_001080547.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010733.hg.1 | NM_001080547.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010733.hg.1 | NM_001080547.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010733.hg.1 | NM_001080547.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010733.hg.1 | NM_001080547.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010733.hg.1 | NM_001080547.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010733.hg.1 | NM_001080547.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010733.hg.1 | NM_001080547.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010733.hg.1 | NM_001080547.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010733.hg.1 | NM_001080547.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010733.hg.1 | NM_001080547.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010733.hg.1 | NM_001080547.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010733.hg.1 | NM_003120.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010733.hg.1 | NM_003120.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010733.hg.1 | NM_003120.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010733.hg.1 | NM_003120.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010733.hg.1 | NM_003120.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010733.hg.1 | NM_003120.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010733.hg.1 | NM_003120.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010733.hg.1 | NM_003120.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010733.hg.1 | NM_003120.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010733.hg.1 | NM_003120.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010733.hg.1 | NM_003120.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010733.hg.1 | NM_003120.2 | Refseq |

SPI1
Genomic Map