Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR15016288.hg.1 |
ENST00000314852.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15016288.hg.1 |
ENST00000403490.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15016288.hg.1 |
ENST00000542473.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15016288.hg.1 |
NM_018285.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15016288.hg.1 |
TC15001669.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR15016289.hg.1 |
ENST00000314852.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15016289.hg.1 |
ENST00000403490.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15016289.hg.1 |
NM_018285.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15016289.hg.1 |
TC15001669.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR15016290.hg.1 |
ENST00000314852.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15016290.hg.1 |
ENST00000403490.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15016290.hg.1 |
ENST00000542473.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15016290.hg.1 |
NM_018285.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15016290.hg.1 |
TC15001669.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR15016293.hg.1 |
ENST00000314852.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15016293.hg.1 |
NM_018285.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15016293.hg.1 |
TC15001669.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010080.hg.1 |
ENST00000314852.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010080.hg.1 |
ENST00000314852.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010080.hg.1 |
ENST00000314852.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010080.hg.1 |
ENST00000314852.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010080.hg.1 |
ENST00000314852.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010080.hg.1 |
ENST00000314852.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010080.hg.1 |
ENST00000314852.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010080.hg.1 |
ENST00000314852.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010080.hg.1 |
ENST00000403490.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010080.hg.1 |
ENST00000403490.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010080.hg.1 |
ENST00000403490.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010080.hg.1 |
ENST00000403490.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010080.hg.1 |
ENST00000403490.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010080.hg.1 |
ENST00000403490.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010080.hg.1 |
ENST00000565349.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010080.hg.1 |
ENST00000565349.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010080.hg.1 |
ENST00000565349.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010080.hg.1 |
NM_018285.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010080.hg.1 |
NM_018285.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010080.hg.1 |
NM_018285.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010080.hg.1 |
NM_018285.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010080.hg.1 |
NM_018285.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010080.hg.1 |
NM_018285.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009528.hg.1 |
ENST00000427532.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15016294.hg.1 |
NM_018285.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15016294.hg.1 |
TC15001669.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1500175375.hg.1 |
ENST00000314852.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500175375.hg.1 |
ENST00000403490.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500175375.hg.1 |
NM_018285.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500175375.hg.1 |
ENST00000314852.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500175375.hg.1 |
ENST00000403490.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500175375.hg.1 |
NM_018285.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500175375.hg.1 |
TC1500010080.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1500175378.hg.1 |
ENST00000403490.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500175378.hg.1 |
NM_018285.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500175378.hg.1 |
ENST00000403490.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500175378.hg.1 |
NM_018285.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500175378.hg.1 |
TC1500010080.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1500175377.hg.1 |
ENST00000314852.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500175377.hg.1 |
ENST00000403490.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500175377.hg.1 |
NM_018285.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500175377.hg.1 |
ENST00000314852.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500175377.hg.1 |
ENST00000403490.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500175377.hg.1 |
NM_018285.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500175377.hg.1 |
TC1500010080.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0900009528.hg.1 |
ENST00000427532.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010080.hg.1 |
ENST00000314852.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010080.hg.1 |
ENST00000314852.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010080.hg.1 |
ENST00000314852.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010080.hg.1 |
ENST00000314852.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010080.hg.1 |
ENST00000314852.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010080.hg.1 |
ENST00000314852.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010080.hg.1 |
ENST00000314852.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010080.hg.1 |
ENST00000314852.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010080.hg.1 |
ENST00000403490.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010080.hg.1 |
ENST00000403490.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010080.hg.1 |
ENST00000403490.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010080.hg.1 |
ENST00000403490.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010080.hg.1 |
ENST00000403490.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010080.hg.1 |
ENST00000403490.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010080.hg.1 |
ENST00000565349.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010080.hg.1 |
ENST00000565349.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010080.hg.1 |
ENST00000565349.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010080.hg.1 |
NM_018285.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010080.hg.1 |
NM_018285.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010080.hg.1 |
NM_018285.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010080.hg.1 |
NM_018285.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010080.hg.1 |
NM_018285.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010080.hg.1 |
NM_018285.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500175374.hg.1 |
ENST00000314852.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500175374.hg.1 |
ENST00000403490.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500175374.hg.1 |
NM_018285.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500175374.hg.1 |
ENST00000314852.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500175374.hg.1 |
ENST00000403490.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500175374.hg.1 |
NM_018285.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500175374.hg.1 |
TC1500010080.hg.1 |
Clariom D TC |