Assay Details
| Gene Symbol: YAF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190979.2 | NP_001177908.1 | 1 - 2 | 214 | 81 | |
| NM_001190980.2 | NP_001177909.1 | 1 - 2 | 214 | 81 | ||
| NM_001320080.1 | NP_001307009.1 | 1 - 2 | 214 | 81 | ||
| NM_005748.5 | NP_005739.2 | 1 - 2 | 214 | 81 | ||
| NR_135139.1 | 1 - 2 | 214 | 81 | |||
| NR_135140.1 | 1 - 2 | 214 | 81 | |||
| XM_011537728.2 | XP_011536030.1 | 1 - 2 | 139 | 81 | ||
| XM_017018670.1 | XP_016874159.1 | 1 - 2 | 152 | 81 | ||
| GenBank mRNA | AK127531.1 | - | 1 - 2 | 94 | 81 | |
| AK291355.1 | - | 1 - 2 | 94 | 81 | ||
| AK296191.1 | - | 1 - 2 | 94 | 81 | ||
| BC037777.2 | - | 1 - 2 | 82 | 5276663 | 81 | |
| CB963485.1 | - | 1 - 2 | 69 | 81 | ||
| CB993159.1 | - | 1 - 2 | 93 | 81 | ||
| DA011078.1 | - | 1 - 2 | 195 | 81 | ||
| DA491895.1 | - | 1 - 2 | 198 | 81 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK291355.1, AK296191.1, BC037777.2, NM_001190979.2, NM_001190980.2, NM_001320080.1, NM_005748.5, XM_011537728.2, XM_017018670.1 transcripts.
Set Membership: |
|

YAF2
Genomic Map