Assay Details
| Gene Symbol: PRPF40B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031698.2 | NP_001026868.2 | 6 - 7 | 530 | 105 | |
| NM_012272.2 | NP_036404.1 | 5 - 6 | 820 | 105 | ||
| XM_006719324.3 | XP_006719387.1 | 5 - 6 | 1584 | 77 | ||
| XM_006719325.3 | XP_006719388.1 | 5 - 6 | 1584 | 77 | ||
| XM_011538135.2 | XP_011536437.1 | 5 - 6 | 1584 | 77 | ||
| XM_011538136.2 | XP_011536438.1 | 5 - 6 | 1584 | 77 | ||
| XM_011538137.2 | XP_011536439.1 | 4 - 5 | 1503 | 77 | ||
| XM_011538139.2 | XP_011536441.1 | 6 - 7 | 524 | 105 | ||
| XM_011538140.2 | XP_011536442.1 | 6 - 7 | 524 | 105 | ||
| XM_011538141.1 | XP_011536443.1 | 5 - 6 | 350 | 105 | ||
| XM_011538143.2 | XP_011536445.1 | 5 - 6 | 366 | 105 | ||
| XM_017019135.1 | XP_016874624.1 | 5 - 6 | 287 | 105 | ||
| XM_017019138.1 | XP_016874627.1 | 5 - 6 | 1584 | 77 | ||
| GenBank mRNA | AF049525.1 | - | 4 - 5 | 184 | 105 | |
| AL834216.1 | - | 5 - 6 | 820 | 105 | ||
| BC031917.1 | - | 3 - 4 | 126 | 105 | ||
| BC050398.2 | - | 6 - 7 | 366 | 105 | ||
| BC067364.1 | - | 5 - 6 | 351 | 6644672 | 105 | |
More Information
Set Membership: |
|

PRPF40B
Genomic Map