Assay Details
| Gene Symbol: SLC35A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032289.2 | NP_001027460.1 | 3 - 4 | 749 | 82 | |
| NM_001282647.1 | NP_001269576.1 | 3 - 4 | 749 | 82 | ||
| NM_001282648.1 | NP_001269577.1 | 3 - 4 | 436 | 82 | ||
| GenBank mRNA | AK290284.1 | - | 3 - 4 | 437 | 82 | |
| AL556714.3 | - | 3 - 4 | 436 | 82 | ||
| BC035747.1 | - | 3 - 4 | 447 | 5749525 | 82 | |
| BI820134.1 | - | 3 - 4 | 420 | 82 | ||
More Information
Set Membership: |
|

SLC35A2
Genomic Map