Assay Details
| Gene Symbol: XIAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001167.3 | NP_001158.2 | 6 - 7 | 1447 | 98 | |
| NM_001204401.1 | NP_001191330.1 | 6 - 7 | 1414 | 98 | ||
| NR_037916.1 | 5 - 6 | 538 | 98 | |||
| XM_006724754.2 | XP_006724817.1 | 7 - 8 | 1527 | 98 | ||
| XM_011531329.2 | XP_011529631.1 | 6 - 7 | 1417 | 98 | ||
| GenBank mRNA | AK309704.1 | - | 5 - 6 | 509 | 98 | |
| AK313883.1 | - | 6 - 7 | 1414 | 98 | ||
| BC032729.1 | - | 6 - 7 | 1366 | 5532247 | 98 | |
| U32974.1 | - | 6 - 7 | 1369 | 98 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

XIAP
Genomic Map