Assay Details
Gene Symbol: FGF12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_004113.5 | NP_004104.3 | 1 - 2 | 118 | 59 | |
GenBank mRNA | AK312513.1 | - | 1 - 2 | 75 | 59 | |
BC022524.1 | - | 1 - 2 | 102 | 4794600 | 59 | |
DA521146.1 | - | 1 - 2 | 118 | 59 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK312513.1, BC022524.1, NM_004113.5 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
regulation of membrane depolarization
signal transduction
cell-cell signaling
chemical synaptic transmission
nervous system development
heart development
adult locomotory behavior
fibroblast growth factor receptor signaling pathway
positive regulation of sodium ion transport
neuromuscular process
cardiac muscle cell action potential involved in contraction
regulation of neuronal action potential
regulation of sodium ion transmembrane transport
regulation of voltage-gated sodium channel activity
regulation of sodium ion transmembrane transporter activity
negative regulation of cation channel activity
protein binding
growth factor activity
sodium channel regulator activity
ion channel binding
regulation of membrane depolarization
signal transduction
cell-cell signaling
chemical synaptic transmission
nervous system development
heart development
adult locomotory behavior
fibroblast growth factor receptor signaling pathway
positive regulation of sodium ion transport
neuromuscular process
cardiac muscle cell action potential involved in contraction
regulation of neuronal action potential
regulation of sodium ion transmembrane transport
regulation of voltage-gated sodium channel activity
regulation of sodium ion transmembrane transporter activity
negative regulation of cation channel activity
protein binding
growth factor activity
sodium channel regulator activity
ion channel binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0300013547.hg.1 | ENST00000363314.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013547.hg.1 | ENST00000418610.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013547.hg.1 | ENST00000418610.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013547.hg.1 | ENST00000418610.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013547.hg.1 | ENST00000418610.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013547.hg.1 | ENST00000418610.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013547.hg.1 | ENST00000418610.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013547.hg.1 | ENST00000430714.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013547.hg.1 | ENST00000430714.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013547.hg.1 | ENST00000430714.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013547.hg.1 | ENST00000430714.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013547.hg.1 | ENST00000430714.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013547.hg.1 | ENST00000430714.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013547.hg.1 | ENST00000440901.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013547.hg.1 | ENST00000440901.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013547.hg.1 | ENST00000440901.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013547.hg.1 | ENST00000440901.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013547.hg.1 | ENST00000440901.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013547.hg.1 | ENST00000440901.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013547.hg.1 | ENST00000445105.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013547.hg.1 | ENST00000445105.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013547.hg.1 | ENST00000445105.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013547.hg.1 | ENST00000445105.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013547.hg.1 | ENST00000445105.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013547.hg.1 | ENST00000445105.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013547.hg.1 | ENST00000445105.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013547.hg.1 | ENST00000445105.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013547.hg.1 | ENST00000445105.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013547.hg.1 | ENST00000445105.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013547.hg.1 | ENST00000445105.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013547.hg.1 | ENST00000445105.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013547.hg.1 | ENST00000445105.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013547.hg.1 | ENST00000448795.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013547.hg.1 | ENST00000448795.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013547.hg.1 | ENST00000448795.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013547.hg.1 | ENST00000448795.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013547.hg.1 | ENST00000448795.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013547.hg.1 | ENST00000448795.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013547.hg.1 | ENST00000450716.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013547.hg.1 | ENST00000450716.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013547.hg.1 | ENST00000450716.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013547.hg.1 | ENST00000450716.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013547.hg.1 | ENST00000450716.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013547.hg.1 | ENST00000450716.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013547.hg.1 | ENST00000450716.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013547.hg.1 | ENST00000450716.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013547.hg.1 | ENST00000454309.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013547.hg.1 | ENST00000454309.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013547.hg.1 | ENST00000454309.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013547.hg.1 | ENST00000454309.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013547.hg.1 | ENST00000454309.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013547.hg.1 | ENST00000454309.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013547.hg.1 | ENST00000454309.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013547.hg.1 | ENST00000454309.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013547.hg.1 | ENST00000454309.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013547.hg.1 | ENST00000454309.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013547.hg.1 | ENST00000466144.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013547.hg.1 | ENST00000466144.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013547.hg.1 | ENST00000466144.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013547.hg.1 | ENST00000466144.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013547.hg.1 | ENST00000491636.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013547.hg.1 | ENST00000491636.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013547.hg.1 | ENST00000491636.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013547.hg.1 | ENST00000491636.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013547.hg.1 | NM_004113.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013547.hg.1 | NM_004113.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013547.hg.1 | NM_004113.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013547.hg.1 | NM_004113.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013547.hg.1 | NM_004113.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013547.hg.1 | NM_004113.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013547.hg.1 | NM_004113.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013547.hg.1 | NM_004113.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013547.hg.1 | NM_004113.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013547.hg.1 | NM_004113.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013547.hg.1 | NM_004113.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013547.hg.1 | NM_004113.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013547.hg.1 | NM_004113.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013547.hg.1 | NM_021032.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013547.hg.1 | NM_021032.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013547.hg.1 | NM_021032.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013547.hg.1 | NM_021032.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013547.hg.1 | NM_021032.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013547.hg.1 | NM_021032.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013547.hg.1 | NM_021032.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013547.hg.1 | NM_021032.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013547.hg.1 | NM_021032.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013547.hg.1 | NM_021032.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013547.hg.1 | NM_021032.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013547.hg.1 | NM_021032.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013547.hg.1 | ENST00000363314.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013547.hg.1 | ENST00000418610.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013547.hg.1 | ENST00000418610.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013547.hg.1 | ENST00000418610.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013547.hg.1 | ENST00000418610.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013547.hg.1 | ENST00000418610.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013547.hg.1 | ENST00000418610.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013547.hg.1 | ENST00000430714.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013547.hg.1 | ENST00000430714.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013547.hg.1 | ENST00000430714.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013547.hg.1 | ENST00000430714.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013547.hg.1 | ENST00000430714.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013547.hg.1 | ENST00000430714.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013547.hg.1 | ENST00000440901.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013547.hg.1 | ENST00000440901.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013547.hg.1 | ENST00000440901.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013547.hg.1 | ENST00000440901.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013547.hg.1 | ENST00000440901.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013547.hg.1 | ENST00000440901.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013547.hg.1 | ENST00000445105.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013547.hg.1 | ENST00000445105.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013547.hg.1 | ENST00000445105.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013547.hg.1 | ENST00000445105.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013547.hg.1 | ENST00000445105.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013547.hg.1 | ENST00000445105.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013547.hg.1 | ENST00000445105.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013547.hg.1 | ENST00000445105.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013547.hg.1 | ENST00000445105.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013547.hg.1 | ENST00000445105.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013547.hg.1 | ENST00000445105.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013547.hg.1 | ENST00000445105.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013547.hg.1 | ENST00000445105.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013547.hg.1 | ENST00000448795.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013547.hg.1 | ENST00000448795.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013547.hg.1 | ENST00000448795.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013547.hg.1 | ENST00000448795.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013547.hg.1 | ENST00000448795.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013547.hg.1 | ENST00000448795.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013547.hg.1 | ENST00000450716.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013547.hg.1 | ENST00000450716.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013547.hg.1 | ENST00000450716.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013547.hg.1 | ENST00000450716.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013547.hg.1 | ENST00000450716.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013547.hg.1 | ENST00000450716.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013547.hg.1 | ENST00000450716.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013547.hg.1 | ENST00000450716.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013547.hg.1 | ENST00000454309.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013547.hg.1 | ENST00000454309.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013547.hg.1 | ENST00000454309.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013547.hg.1 | ENST00000454309.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013547.hg.1 | ENST00000454309.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013547.hg.1 | ENST00000454309.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013547.hg.1 | ENST00000454309.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013547.hg.1 | ENST00000454309.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013547.hg.1 | ENST00000454309.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013547.hg.1 | ENST00000454309.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013547.hg.1 | ENST00000466144.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013547.hg.1 | ENST00000466144.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013547.hg.1 | ENST00000466144.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013547.hg.1 | ENST00000466144.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013547.hg.1 | ENST00000491636.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013547.hg.1 | ENST00000491636.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013547.hg.1 | ENST00000491636.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013547.hg.1 | ENST00000491636.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013547.hg.1 | NM_004113.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013547.hg.1 | NM_004113.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013547.hg.1 | NM_004113.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013547.hg.1 | NM_004113.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013547.hg.1 | NM_004113.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013547.hg.1 | NM_004113.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013547.hg.1 | NM_004113.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013547.hg.1 | NM_004113.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013547.hg.1 | NM_004113.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013547.hg.1 | NM_004113.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013547.hg.1 | NM_004113.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013547.hg.1 | NM_004113.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013547.hg.1 | NM_004113.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013547.hg.1 | NM_021032.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013547.hg.1 | NM_021032.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013547.hg.1 | NM_021032.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013547.hg.1 | NM_021032.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013547.hg.1 | NM_021032.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013547.hg.1 | NM_021032.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013547.hg.1 | NM_021032.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013547.hg.1 | NM_021032.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013547.hg.1 | NM_021032.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013547.hg.1 | NM_021032.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013547.hg.1 | NM_021032.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013547.hg.1 | NM_021032.4 | Refseq |
