Assay Details
| Gene Symbol: WWOX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016373.3 | NP_057457.1 | 1 - 2 | 475 | 94 | |
| NM_130791.3 | NP_570607.1 | 1 - 2 | 475 | 94 | ||
| XM_011523101.2 | XP_011521403.1 | 1 - 2 | 234 | 94 | ||
| XM_011523103.2 | XP_011521405.1 | 1 - 2 | 234 | 94 | ||
| XM_011523104.2 | XP_011521406.1 | 1 - 2 | 234 | 94 | ||
| XM_011523105.2 | XP_011521407.1 | 1 - 2 | 234 | 94 | ||
| XM_017023278.1 | XP_016878767.1 | 1 - 2 | 234 | 94 | ||
| GenBank mRNA | AF187015.1 | - | 1 - 2 | 193 | 94 | |
| AF211943.1 | - | 1 - 2 | 234 | 94 | ||
| AF227526.1 | - | 1 - 2 | 194 | 94 | ||
| AF395123.1 | - | 1 - 2 | 242 | 94 | ||
| AF395124.1 | - | 1 - 2 | 242 | 94 | ||
| AK027626.1 | - | 1 - 2 | 233 | 94 | ||
| AK290438.1 | - | 1 - 2 | 225 | 94 | ||
| AK291300.1 | - | 1 - 2 | 232 | 94 | ||
| AW874692.1 | - | 1 - 2 | 120 | 94 | ||
| AY256821.1 | - | 1 - 2 | 164 | 94 | ||
| BC003184.1 | - | 1 - 2 | 188 | 3535809 | 94 | |
| BT007445.1 | - | 109 | 94 | |||
| DB035405.1 | - | 1 - 2 | 475 | 94 | ||
More Information
Set Membership: |
|

WWOX
Genomic Map