Assay Details
| Gene Symbol: RHEB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_005614.3 | NP_005605.1 | 7 - 8 | 873 | 132 | |
| XM_011516457.1 | XP_011514759.1 | 8 - 9 | 927 | 132 | ||
| GenBank mRNA | AF148645.1 | - | 7 - 8 | 132 | ||
| AF493921.1 | - | 7 - 8 | 460 | 132 | ||
| AK125446.1 | - | 7 - 8 | 871 | 132 | ||
| AK289370.1 | - | 7 - 8 | 602 | 132 | ||
| BC016155.1 | - | 7 - 8 | 873 | 3528583 | 132 | |
| BC066307.1 | - | 1 | 631 | 5274556 | 132 | |
| BC107705.1 | - | 1 | 822 | 6615350 | 132 | |
| BT006958.1 | - | 7 - 8 | 460 | 132 | ||
| D78132.1 | - | 7 - 8 | 477 | 132 | ||
| Z29677.1 | - | 7 - 8 | 483 | 132 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC07002040.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
cell cycle arrest
signal transduction
small GTPase mediated signal transduction
macroautophagy
positive regulation of TOR signaling
positive regulation of oligodendrocyte differentiation
regulation of type B pancreatic cell development
GTPase activity
GTP binding
protein kinase binding
metal ion binding
cell cycle arrest
signal transduction
small GTPase mediated signal transduction
macroautophagy
positive regulation of TOR signaling
positive regulation of oligodendrocyte differentiation
regulation of type B pancreatic cell development
GTPase activity
GTP binding
protein kinase binding
metal ion binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0700013080.hg.1 | ENST00000262187.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013080.hg.1 | ENST00000262187.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013080.hg.1 | ENST00000262187.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013080.hg.1 | ENST00000262187.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013080.hg.1 | ENST00000262187.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013080.hg.1 | ENST00000262187.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013080.hg.1 | ENST00000262187.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013080.hg.1 | ENST00000262187.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013080.hg.1 | ENST00000262187.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013080.hg.1 | ENST00000262187.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013080.hg.1 | ENST00000262187.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013080.hg.1 | ENST00000262187.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013080.hg.1 | ENST00000262187.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013080.hg.1 | ENST00000262187.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013080.hg.1 | ENST00000262187.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013080.hg.1 | ENST00000262187.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013080.hg.1 | ENST00000470072.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013080.hg.1 | ENST00000470072.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013080.hg.1 | ENST00000470072.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013080.hg.1 | ENST00000470072.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013080.hg.1 | ENST00000470072.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013080.hg.1 | ENST00000470072.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013080.hg.1 | ENST00000470370.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013080.hg.1 | ENST00000470370.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013080.hg.1 | ENST00000470370.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013080.hg.1 | ENST00000470370.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013080.hg.1 | ENST00000470370.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013080.hg.1 | ENST00000470370.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013080.hg.1 | ENST00000472642.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013080.hg.1 | ENST00000472642.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013080.hg.1 | ENST00000472642.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013080.hg.1 | ENST00000472642.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013080.hg.1 | ENST00000472642.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013080.hg.1 | ENST00000472642.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013080.hg.1 | ENST00000472642.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013080.hg.1 | ENST00000472642.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013080.hg.1 | ENST00000472642.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013080.hg.1 | ENST00000472642.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013080.hg.1 | ENST00000478470.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013080.hg.1 | ENST00000478470.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013080.hg.1 | ENST00000478470.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013080.hg.1 | ENST00000478470.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013080.hg.1 | ENST00000478470.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013080.hg.1 | ENST00000478470.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013080.hg.1 | ENST00000478470.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013080.hg.1 | ENST00000478470.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013080.hg.1 | ENST00000478470.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013080.hg.1 | ENST00000478470.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013080.hg.1 | ENST00000478470.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013080.hg.1 | ENST00000478470.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013080.hg.1 | ENST00000482053.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013080.hg.1 | ENST00000482053.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013080.hg.1 | ENST00000482053.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013080.hg.1 | ENST00000482053.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013080.hg.1 | ENST00000496004.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013080.hg.1 | ENST00000496004.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013080.hg.1 | ENST00000496004.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013080.hg.1 | ENST00000496004.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013080.hg.1 | ENST00000496004.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013080.hg.1 | ENST00000496004.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013080.hg.1 | ENST00000496004.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013080.hg.1 | ENST00000496004.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013080.hg.1 | ENST00000496004.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013080.hg.1 | NM_005614.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013080.hg.1 | NM_005614.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013080.hg.1 | NM_005614.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013080.hg.1 | NM_005614.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013080.hg.1 | NM_005614.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013080.hg.1 | NM_005614.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013080.hg.1 | NM_005614.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013080.hg.1 | NM_005614.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013080.hg.1 | NM_005614.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013080.hg.1 | NM_005614.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013080.hg.1 | NM_005614.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013080.hg.1 | NM_005614.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013080.hg.1 | NM_005614.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013080.hg.1 | NM_005614.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013080.hg.1 | NM_005614.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013080.hg.1 | NM_005614.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07031249.hg.1 | TC07002040.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07031249.hg.1 | ENST00000262187.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07031249.hg.1 | ENST00000472642.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07031249.hg.1 | ENST00000496004.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07031249.hg.1 | NM_005614.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700194414.hg.1 | ENST00000262187.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700194414.hg.1 | ENST00000472642.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700194414.hg.1 | ENST00000478470.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700194414.hg.1 | ENST00000496004.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700194414.hg.1 | NM_005614.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700194414.hg.1 | ENST00000262187.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700194414.hg.1 | ENST00000472642.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700194414.hg.1 | ENST00000478470.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700194414.hg.1 | ENST00000496004.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700194414.hg.1 | NM_005614.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700194414.hg.1 | TC0700013080.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0700013080.hg.1 | ENST00000262187.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013080.hg.1 | ENST00000262187.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013080.hg.1 | ENST00000262187.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013080.hg.1 | ENST00000262187.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013080.hg.1 | ENST00000262187.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013080.hg.1 | ENST00000262187.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013080.hg.1 | ENST00000262187.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013080.hg.1 | ENST00000262187.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013080.hg.1 | ENST00000262187.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013080.hg.1 | ENST00000262187.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013080.hg.1 | ENST00000262187.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013080.hg.1 | ENST00000262187.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013080.hg.1 | ENST00000262187.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013080.hg.1 | ENST00000262187.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013080.hg.1 | ENST00000262187.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013080.hg.1 | ENST00000262187.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013080.hg.1 | ENST00000470072.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013080.hg.1 | ENST00000470072.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013080.hg.1 | ENST00000470072.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013080.hg.1 | ENST00000470072.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013080.hg.1 | ENST00000470072.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013080.hg.1 | ENST00000470072.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013080.hg.1 | ENST00000470370.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013080.hg.1 | ENST00000470370.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013080.hg.1 | ENST00000470370.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013080.hg.1 | ENST00000470370.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013080.hg.1 | ENST00000470370.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013080.hg.1 | ENST00000470370.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013080.hg.1 | ENST00000472642.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013080.hg.1 | ENST00000472642.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013080.hg.1 | ENST00000472642.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013080.hg.1 | ENST00000472642.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013080.hg.1 | ENST00000472642.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013080.hg.1 | ENST00000472642.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013080.hg.1 | ENST00000472642.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013080.hg.1 | ENST00000472642.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013080.hg.1 | ENST00000472642.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013080.hg.1 | ENST00000472642.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013080.hg.1 | ENST00000478470.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013080.hg.1 | ENST00000478470.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013080.hg.1 | ENST00000478470.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013080.hg.1 | ENST00000478470.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013080.hg.1 | ENST00000478470.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013080.hg.1 | ENST00000478470.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013080.hg.1 | ENST00000478470.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013080.hg.1 | ENST00000478470.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013080.hg.1 | ENST00000478470.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013080.hg.1 | ENST00000478470.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013080.hg.1 | ENST00000478470.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013080.hg.1 | ENST00000478470.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013080.hg.1 | ENST00000482053.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013080.hg.1 | ENST00000482053.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013080.hg.1 | ENST00000482053.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013080.hg.1 | ENST00000482053.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013080.hg.1 | ENST00000496004.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013080.hg.1 | ENST00000496004.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013080.hg.1 | ENST00000496004.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013080.hg.1 | ENST00000496004.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013080.hg.1 | ENST00000496004.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013080.hg.1 | ENST00000496004.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013080.hg.1 | ENST00000496004.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013080.hg.1 | ENST00000496004.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013080.hg.1 | ENST00000496004.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013080.hg.1 | NM_005614.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013080.hg.1 | NM_005614.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013080.hg.1 | NM_005614.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013080.hg.1 | NM_005614.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013080.hg.1 | NM_005614.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013080.hg.1 | NM_005614.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013080.hg.1 | NM_005614.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013080.hg.1 | NM_005614.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013080.hg.1 | NM_005614.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013080.hg.1 | NM_005614.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013080.hg.1 | NM_005614.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013080.hg.1 | NM_005614.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013080.hg.1 | NM_005614.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013080.hg.1 | NM_005614.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013080.hg.1 | NM_005614.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013080.hg.1 | NM_005614.3 | Refseq |

RHEB
Genomic Map