Assay Details
| Gene Symbol: CACFD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001135775.2 | NP_001129247.1 | 3 - 4 | 403 | 77 | |
| NM_017586.3 | NP_060056.1 | 4 - 5 | 529 | 77 | ||
| XM_011518185.2 | XP_011516487.1 | 4 - 5 | 2086 | 77 | ||
| XM_011518186.1 | XP_011516488.1 | 4 - 5 | 582 | 77 | ||
| GenBank mRNA | AJ011373.1 | - | 4 - 5 | 485 | 77 | |
| AK074852.1 | - | 4 - 5 | 507 | 77 | ||
| AK075416.1 | - | 3 - 4 | 401 | 77 | ||
| AK075548.1 | - | 4 - 5 | 497 | 77 | ||
| AK304524.1 | - | 5 - 6 | 487 | 77 | ||
| AK311325.1 | - | 4 - 5 | 585 | 77 | ||
| AY358452.1 | - | 4 - 5 | 477 | 77 | ||
| BC030558.1 | - | 4 - 5 | 495 | 5223979 | 77 | |
| DA699497.1 | - | 3 - 4 | 340 | 77 | ||
| DB165724.1 | - | 4 - 5 | 529 | 77 | ||
More Information
Set Membership: |
|

CACFD1
Genomic Map