Assay Details
Gene Symbol: CNIH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_182553.2 | NP_872359.1 | 2 - 3 | 406 | 85 | |
NR_073079.1 | 2 - 3 | 402 | 85 | |||
GenBank mRNA | BC047953.1 | - | 2 - 3 | 382 | 85 | |
BM547951.1 | - | 406 | 85 | |||
DR760677.1 | - | 2 - 3 | 378 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
