Assay Details
Gene Symbol: TAC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077503.1 | NP_001070971.1 | 2 - 3 | 195 | 66 | |
GenBank mRNA | AF515829.1 | - | 2 - 3 | 195 | 66 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC17001673.hg.1
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | ENST00000326219.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | ENST00000326219.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | ENST00000326219.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | ENST00000326219.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | ENST00000326219.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | ENST00000334568.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | ENST00000334568.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | ENST00000334568.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | ENST00000334568.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | ENST00000334568.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | ENST00000334568.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | ENST00000352793.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | ENST00000352793.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | ENST00000352793.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | ENST00000352793.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | ENST00000398154.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | ENST00000398154.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | ENST00000398154.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | ENST00000398154.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | ENST00000398154.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | ENST00000436235.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | ENST00000436235.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | ENST00000436235.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | ENST00000436235.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | ENST00000436235.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | ENST00000436235.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | ENST00000503603.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | ENST00000503603.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | ENST00000608380.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | ENST00000608380.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | ENST00000608861.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | ENST00000608861.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | NM_001077503.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | NM_001077503.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | NM_001077503.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | NM_001077503.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | NM_001077503.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | NM_001077504.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | NM_001077504.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | NM_001077504.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | NM_001077504.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | NM_001077504.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | NM_001077505.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | NM_001077505.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | NM_001077505.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | NM_001077505.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | NM_001077506.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | NM_001077506.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | NM_001077506.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | NM_001077506.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | NM_001077506.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | NM_001077506.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | NM_170685.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | NM_170685.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | NM_170685.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | NM_170685.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | NM_170685.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011066.hg.1 | NM_170685.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17022501.hg.1 | ENST00000334568.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17022501.hg.1 | ENST00000398154.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17022501.hg.1 | ENST00000436235.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17022501.hg.1 | NM_001077503.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17022501.hg.1 | NM_001077506.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17022501.hg.1 | NM_170685.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17022501.hg.1 | TC17001673.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1700189059.hg.1 | ENST00000334568.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700189059.hg.1 | ENST00000398154.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700189059.hg.1 | ENST00000436235.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700189059.hg.1 | NM_001077503.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700189059.hg.1 | NM_001077506.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700189059.hg.1 | NM_170685.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700189059.hg.1 | ENST00000334568.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700189059.hg.1 | ENST00000398154.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700189059.hg.1 | ENST00000436235.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700189059.hg.1 | NM_001077503.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700189059.hg.1 | NM_001077506.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700189059.hg.1 | NM_170685.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700189059.hg.1 | TC1700011066.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | ENST00000326219.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | ENST00000326219.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | ENST00000326219.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | ENST00000326219.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | ENST00000326219.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | ENST00000334568.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | ENST00000334568.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | ENST00000334568.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | ENST00000334568.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | ENST00000334568.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | ENST00000334568.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | ENST00000352793.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | ENST00000352793.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | ENST00000352793.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | ENST00000352793.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | ENST00000398154.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | ENST00000398154.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | ENST00000398154.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | ENST00000398154.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | ENST00000398154.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | ENST00000436235.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | ENST00000436235.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | ENST00000436235.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | ENST00000436235.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | ENST00000436235.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | ENST00000436235.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | ENST00000503603.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | ENST00000503603.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | ENST00000608380.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | ENST00000608380.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | ENST00000608861.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | ENST00000608861.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | NM_001077503.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | NM_001077503.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | NM_001077503.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | NM_001077503.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | NM_001077503.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | NM_001077504.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | NM_001077504.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | NM_001077504.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | NM_001077504.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | NM_001077504.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | NM_001077505.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | NM_001077505.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | NM_001077505.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | NM_001077505.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | NM_001077506.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | NM_001077506.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | NM_001077506.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | NM_001077506.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | NM_001077506.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | NM_001077506.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | NM_170685.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | NM_170685.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | NM_170685.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | NM_170685.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | NM_170685.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011066.hg.1 | NM_170685.2 | Refseq |
