Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01000347.hg.1 |
TC01000031.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01000347.hg.1 |
ENST00000379198.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01000347.hg.1 |
NM_080605.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006494.hg.1 |
ENST00000379198.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006494.hg.1 |
ENST00000379198.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006494.hg.1 |
NM_080605.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006494.hg.1 |
NM_080605.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01000346.hg.1 |
TC01000031.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01000346.hg.1 |
ENST00000379198.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01000346.hg.1 |
NM_080605.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006494.hg.1 |
ENST00000379198.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006494.hg.1 |
ENST00000379198.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006494.hg.1 |
NM_080605.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006494.hg.1 |
NM_080605.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100144663.hg.1 |
ENST00000379198.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100144663.hg.1 |
NM_080605.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100144663.hg.1 |
ENST00000379198.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100144663.hg.1 |
NM_080605.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100144663.hg.1 |
TC0100006494.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100144661.hg.1 |
ENST00000379198.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100144661.hg.1 |
NM_080605.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100144661.hg.1 |
ENST00000379198.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100144661.hg.1 |
NM_080605.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100144661.hg.1 |
TC0100006494.hg.1 |
Clariom D TC |