Assay Details
| Gene Symbol: FAM101B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_182705.2 | NP_874364.1 | 1 - 2 | 379 | 68 | |
| GenBank mRNA | BC031341.1 | - | 409 | 4908279 | 68 | |
More Information
Set Membership: |
|

FAM101B
Genomic Map