Assay Details
| Gene Symbol: ACTB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101.3 | NP_001092.1 | 1 | 54 | 96 | |
| GenBank mRNA | AK025375.1 | - | 1 | 54 | 96 | |
| AK058019.1 | - | 1 | 54 | 96 | ||
| AK098751.1 | - | 1 | 54 | 96 | ||
| AK130062.1 | - | 1 | 54 | 96 | ||
| AK130157.1 | - | 1 | 54 | 96 | ||
| AK222925.1 | - | 1 | 54 | 96 | ||
| AK223055.1 | - | 1 | 54 | 96 | ||
| AK225414.1 | - | 1 | 54 | 96 | ||
| AK301372.1 | - | 1 | 54 | 96 | ||
| AK304552.1 | - | 1 | 54 | 96 | ||
| AK308277.1 | - | 1 | 54 | 96 | ||
| AK309997.1 | - | 1 | 54 | 96 | ||
| BC001301.1 | - | 1 | 37 | 3451917 | 96 | |
| BC002409.2 | - | 1 | 41 | 2961617 | 96 | |
| BC004251.1 | - | 1 | 31 | 3623067 | 96 | |
| BC013380.2 | - | 1 | 45 | 4025526 | 96 | |
| BC013835.1 | - | 1 | 42 | 96 | ||
| BC014861.1 | - | 1 | 44 | 3870487 | 96 | |
| BC016045.1 | - | 1 | 45 | 4564532 | 96 | |
| X63432.1 | - | 1 | 43 | 96 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK301372.1, AK304552.1, BC001301.1, BC002409.2, BC004251.1, BC013380.2, BC014861.1, BC016045.1, NM_001101.3, X63432.1 transcripts.
Set Membership: |
|

ACTB
Genomic Map