Assay Details
| Gene Symbol: MYL4-HBG2 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF116676.1 | - | 862 | 86 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MYL4-HBG2 fusion| Gene Symbol: MYL4-HBG2 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF116676.1 | - | 862 | 86 | ||